Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94903070-94903303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN3
Entrez Gene ID
1266
Gene Name
calponin 3
Gene Aliases
-
UniGene
Hs.483454
Gene Chromosome Location
Chr.1: 94896949-94927223 on Build GRCh38
Transcripts
NM_001286055.1 NM_001286056.1 NM_001839.4 XM_017000245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3866296 c.227C>T p.S76F Chr.1: 94903141-94903141 on Build GRCh38
4673120 c.220A>T p.N74Y Chr.1: 94903148-94903148 on Build GRCh38
426876 c.185T>G p.I62R Chr.1: 94903183-94903183 on Build GRCh38
6274790 c.199C>A p.P67T Chr.1: 94903169-94903169 on Build GRCh38
4010741 c.222C>T p.N74N Chr.1: 94903146-94903146 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374270794 Chr.1: 94903146-94903146 on Build GRCh38
rs748760285 Chr.1: 94903236-94903236 on Build GRCh38
rs781734730 Chr.1: 94903211-94903211 on Build GRCh38
rs763897729 Chr.1: 94903152-94903154 on Build GRCh38
rs761277986 Chr.1: 94903142-94903142 on Build GRCh38
rs949365414 Chr.1: 94903243-94903243 on Build GRCh38
rs980733818 Chr.1: 94903267-94903267 on Build GRCh38
rs750364418 Chr.1: 94903149-94903149 on Build GRCh38
rs150613685 Chr.1: 94903182-94903182 on Build GRCh38
rs747066574 Chr.1: 94903204-94903204 on Build GRCh38
rs763718705 Chr.1: 94903105-94903105 on Build GRCh38
rs150322247 Chr.1: 94903228-94903228 on Build GRCh38
rs375031488 Chr.1: 94903207-94903207 on Build GRCh38
rs917961150 Chr.1: 94903242-94903242 on Build GRCh38
rs762652614 Chr.1: 94903101-94903101 on Build GRCh38
rs755847189 Chr.1: 94903152-94903152 on Build GRCh38
rs74832463 Chr.1: 94903118-94903118 on Build GRCh38
rs202195002 Chr.1: 94903106-94903106 on Build GRCh38
rs200254057 Chr.1: 94903222-94903222 on Build GRCh38
rs777910660 Chr.1: 94903192-94903192 on Build GRCh38
rs1032906772 Chr.1: 94903145-94903145 on Build GRCh38
rs753553608 Chr.1: 94903196-94903197 on Build GRCh38
rs758618767 Chr.1: 94903169-94903169 on Build GRCh38
rs573912790 Chr.1: 94903269-94903269 on Build GRCh38
rs201766980 Chr.1: 94903153-94903153 on Build GRCh38
rs753645792 Chr.1: 94903162-94903162 on Build GRCh38

View Full Product Details