Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155951630-155951855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
UniGene
Hs.743352
Gene Chromosome Location
Chr.1: 155946839-155991263 on Build GRCh38
Transcripts
NM_001162383.1 NM_001162384.1 NM_004723.3 XM_005245587.2 XM_005245588.2 XM_005245589.4 XM_005245590.3 XM_005245591.3 XM_005245592.2 XM_005245593.4 XM_005245594.2 XM_006711622.3 XM_006711623.3 XM_006711624.3 XM_006711625.3 XM_006711626.2 XM_011510136.2 XM_011510137.2 XM_011510139.2 XM_011510141.2 XM_011510143.2 XM_017002794.1 XM_017002795.1 XM_017002796.1 XM_017002797.1 XM_017002798.1 XM_017002799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900996 c.2201C>T p.S734L Chr.1: 155951751-155951751 on Build GRCh38
3418134 c.2176C>T p.R726* Chr.1: 155951773-155951773 on Build GRCh38
5900995 c.2114C>T p.S705L Chr.1: 155951751-155951751 on Build GRCh38
6494278 c.2205G>T p.P735P Chr.1: 155951747-155951747 on Build GRCh38
144411 c.2089-1G>C p.? Chr.1: 155951777-155951777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369802368 Chr.1: 155951827-155951827 on Build GRCh38
rs768686704 Chr.1: 155951748-155951748 on Build GRCh38
rs976272819 Chr.1: 155951743-155951743 on Build GRCh38
rs756960799 Chr.1: 155951720-155951720 on Build GRCh38
rs888235804 Chr.1: 155951677-155951677 on Build GRCh38
rs763632020 Chr.1: 155951711-155951711 on Build GRCh38
rs141390530 Chr.1: 155951752-155951752 on Build GRCh38
rs758172355 Chr.1: 155951738-155951738 on Build GRCh38
rs778067769 Chr.1: 155951770-155951770 on Build GRCh38
rs765628017 Chr.1: 155951729-155951729 on Build GRCh38
rs372676156 Chr.1: 155951831-155951831 on Build GRCh38
rs749370225 Chr.1: 155951773-155951773 on Build GRCh38
rs1027147717 Chr.1: 155951663-155951663 on Build GRCh38
rs988864252 Chr.1: 155951781-155951781 on Build GRCh38
rs750720900 Chr.1: 155951797-155951797 on Build GRCh38
rs753339401 Chr.1: 155951716-155951716 on Build GRCh38
rs1007029232 Chr.1: 155951675-155951675 on Build GRCh38
rs369956173 Chr.1: 155951686-155951686 on Build GRCh38
rs1030545033 Chr.1: 155951744-155951744 on Build GRCh38
rs771059240 Chr.1: 155951808-155951808 on Build GRCh38
rs759837753 Chr.1: 155951813-155951813 on Build GRCh38
rs759298692 Chr.1: 155951786-155951786 on Build GRCh38
rs774588626 Chr.1: 155951809-155951809 on Build GRCh38
rs368080078 Chr.1: 155951700-155951700 on Build GRCh38
rs979693922 Chr.1: 155951793-155951793 on Build GRCh38
rs772532992 Chr.1: 155951818-155951818 on Build GRCh38
rs77345976 Chr.1: 155951747-155951747 on Build GRCh38
rs753150792 Chr.1: 155951691-155951691 on Build GRCh38
rs375598663 Chr.1: 155951734-155951734 on Build GRCh38
rs756491839 Chr.1: 155951804-155951804 on Build GRCh38
rs372353830 Chr.1: 155951728-155951728 on Build GRCh38

View Full Product Details