Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52141073-52141328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB5
Entrez Gene ID
10681
Gene Name
G protein subunit beta 5
Gene Aliases
GB5
UniGene
Hs.155090
Gene Chromosome Location
Chr.15: 52120926-52191368 on Build GRCh38
Transcripts
NM_006578.3 NM_016194.3 XM_011521162.2 XM_011521163.2 XM_017021867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502134 c.569C>T p.S190F Chr.15: 52141198-52141198 on Build GRCh38
6672323 c.522C>T p.Y174Y Chr.15: 52141245-52141245 on Build GRCh38
13673 c.598G>T p.A200S Chr.15: 52141169-52141169 on Build GRCh38
6142726 c.624G>A p.M208I Chr.15: 52141143-52141143 on Build GRCh38
328809 c.583A>T p.T195S Chr.15: 52141184-52141184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779382243 Chr.15: 52141229-52141229 on Build GRCh38
rs768266803 Chr.15: 52141267-52141267 on Build GRCh38
rs759786549 Chr.15: 52141280-52141280 on Build GRCh38
rs201052756 Chr.15: 52141163-52141163 on Build GRCh38
rs973016725 Chr.15: 52141136-52141136 on Build GRCh38
rs767865644 Chr.15: 52141289-52141289 on Build GRCh38
rs201409230 Chr.15: 52141192-52141192 on Build GRCh38
rs928160245 Chr.15: 52141216-52141216 on Build GRCh38
rs17612637 Chr.15: 52141200-52141200 on Build GRCh38
rs767668513 Chr.15: 52141187-52141187 on Build GRCh38
rs746514917 Chr.15: 52141232-52141233 on Build GRCh38
rs777566238 Chr.15: 52141298-52141298 on Build GRCh38
rs373920314 Chr.15: 52141292-52141292 on Build GRCh38
rs147094121 Chr.15: 52141155-52141155 on Build GRCh38
rs769446673 Chr.15: 52141279-52141279 on Build GRCh38
rs377403838 Chr.15: 52141196-52141196 on Build GRCh38
rs138461129 Chr.15: 52141171-52141171 on Build GRCh38
rs369321298 Chr.15: 52141123-52141123 on Build GRCh38
rs770550806 Chr.15: 52141113-52141113 on Build GRCh38
rs776424856 Chr.15: 52141277-52141277 on Build GRCh38
rs745478605 Chr.15: 52141232-52141232 on Build GRCh38
rs776128775 Chr.15: 52141279-52141279 on Build GRCh38
rs575737705 Chr.15: 52141178-52141178 on Build GRCh38
rs775019527 Chr.15: 52141137-52141137 on Build GRCh38
rs761513425 Chr.15: 52141158-52141158 on Build GRCh38
rs749629998 Chr.15: 52141096-52141096 on Build GRCh38
rs746531232 Chr.15: 52141276-52141276 on Build GRCh38
rs765140914 Chr.15: 52141170-52141170 on Build GRCh38
rs754436087 Chr.15: 52141214-52141214 on Build GRCh38
rs768601085 Chr.15: 52141144-52141144 on Build GRCh38
rs757548161 Chr.15: 52141228-52141228 on Build GRCh38
rs760563514 Chr.15: 52141141-52141141 on Build GRCh38
rs984684666 Chr.15: 52141126-52141126 on Build GRCh38
rs745592298 Chr.15: 52141122-52141122 on Build GRCh38
rs759880078 Chr.15: 52141186-52141186 on Build GRCh38
rs200972385 Chr.15: 52141242-52141242 on Build GRCh38
rs372743841 Chr.15: 52141114-52141114 on Build GRCh38
rs779486647 Chr.15: 52141252-52141252 on Build GRCh38

View Full Product Details