Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1456864-1457107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304092 c.859G>A p.D287N Chr.19: 1456895-1456895 on Build GRCh38
6602063 c.902C>T p.S301L Chr.19: 1456938-1456938 on Build GRCh38
226814 c.856C>T p.R286C Chr.19: 1456892-1456892 on Build GRCh38
4140315 c.980C>A p.A327E Chr.19: 1457016-1457016 on Build GRCh38
4646266 c.1035G>A p.M345I Chr.19: 1457071-1457071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746605923 Chr.19: 1456912-1456912 on Build GRCh38
rs769594037 Chr.19: 1456919-1456919 on Build GRCh38
rs762949220 Chr.19: 1457023-1457023 on Build GRCh38
rs749213638 Chr.19: 1456938-1456938 on Build GRCh38
rs942843980 Chr.19: 1456927-1456927 on Build GRCh38
rs777434725 Chr.19: 1457020-1457020 on Build GRCh38
rs745997423 Chr.19: 1457033-1457033 on Build GRCh38
rs770975897 Chr.19: 1456939-1456939 on Build GRCh38
rs749190654 Chr.19: 1457021-1457021 on Build GRCh38
rs951582237 Chr.19: 1456893-1456893 on Build GRCh38
rs927248033 Chr.19: 1457024-1457024 on Build GRCh38
rs768478428 Chr.19: 1456913-1456913 on Build GRCh38
rs773151315 Chr.19: 1456934-1456934 on Build GRCh38
rs1013415718 Chr.19: 1457000-1457000 on Build GRCh38
rs774471899 Chr.19: 1456942-1456942 on Build GRCh38
rs112355929 Chr.19: 1457032-1457032 on Build GRCh38
rs771831923 Chr.19: 1456914-1456914 on Build GRCh38
rs1048120921 Chr.19: 1457059-1457059 on Build GRCh38
rs201709261 Chr.19: 1457007-1457007 on Build GRCh38
rs747823483 Chr.19: 1457009-1457009 on Build GRCh38
rs531286394 Chr.19: 1457029-1457029 on Build GRCh38
rs757722260 Chr.19: 1457002-1457002 on Build GRCh38
rs772162693 Chr.19: 1457052-1457052 on Build GRCh38
rs938566336 Chr.19: 1457027-1457027 on Build GRCh38
rs778394276 Chr.19: 1456890-1456890 on Build GRCh38
rs767768420 Chr.19: 1456945-1456945 on Build GRCh38
rs766601241 Chr.19: 1457034-1457034 on Build GRCh38
rs770840559 Chr.19: 1457022-1457022 on Build GRCh38
rs1026648739 Chr.19: 1456917-1456917 on Build GRCh38
rs751591853 Chr.19: 1457042-1457050 on Build GRCh38
rs369493003 Chr.19: 1456918-1456918 on Build GRCh38
rs574792286 Chr.19: 1456935-1456935 on Build GRCh38
rs61735606 Chr.19: 1456957-1456957 on Build GRCh38
rs143870588 Chr.19: 1457003-1457003 on Build GRCh38
rs542130614 Chr.19: 1456951-1456951 on Build GRCh38
rs759750192 Chr.19: 1456944-1456944 on Build GRCh38
rs200347634 Chr.19: 1456885-1456885 on Build GRCh38
rs1007126374 Chr.19: 1456940-1456940 on Build GRCh38
rs993840196 Chr.19: 1456903-1456903 on Build GRCh38
rs776996014 Chr.19: 1457018-1457029 on Build GRCh38
rs779439124 Chr.19: 1456899-1456899 on Build GRCh38
rs757888187 Chr.19: 1456894-1456894 on Build GRCh38
rs995927729 Chr.19: 1457042-1457042 on Build GRCh38
rs560844801 Chr.19: 1456952-1456952 on Build GRCh38
rs549879354 Chr.19: 1457072-1457072 on Build GRCh38
rs754327827 Chr.19: 1456891-1456891 on Build GRCh38
rs918854913 Chr.19: 1456888-1456888 on Build GRCh38
rs1034874548 Chr.19: 1457011-1457011 on Build GRCh38
rs986975286 Chr.19: 1456905-1456905 on Build GRCh38
rs956778902 Chr.19: 1456976-1456976 on Build GRCh38
rs781037694 Chr.19: 1457008-1457008 on Build GRCh38
rs892389129 Chr.19: 1456969-1456969 on Build GRCh38
rs879174157 Chr.19: 1456916-1456916 on Build GRCh38
rs750529335 Chr.19: 1457014-1457014 on Build GRCh38

View Full Product Details