Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76166558-76166831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH2L
Entrez Gene ID
55668
Gene Name
G-patch domain containing 2 like
Gene Aliases
C14orf118
UniGene
Hs.410231
Gene Chromosome Location
Chr.14: 76151885-76214473 on Build GRCh38
Transcripts
NM_001322026.1 NM_001322027.1 NM_001322028.1 NM_001322029.1 NM_001322030.1 NM_001322031.1 NM_001322032.1 NM_017926.3 NM_017972.3 NR_110314.1 XM_006720191.2 XM_017021427.1 XM_017021428.1 XM_017021429.1 XM_017021430.1 XM_017021431.1 XM_017021432.1 XM_017021433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671975 c.718G>T p.G240W Chr.14: 76166718-76166718 on Build GRCh38
5614163 c.725A>T p.Q242L Chr.14: 76166725-76166725 on Build GRCh38
4747500 c.674_675delGT p.C227fs*1 Chr.14: 76166674-76166675 on Build GRCh38
6673040 c.722G>A p.R241Q Chr.14: 76166722-76166722 on Build GRCh38
3671977 c.718G>T p.G240W Chr.14: 76166718-76166718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769144967 Chr.14: 76166721-76166721 on Build GRCh38
rs117607472 Chr.14: 76166735-76166735 on Build GRCh38
rs776108162 Chr.14: 76166733-76166733 on Build GRCh38
rs765250996 Chr.14: 76166626-76166626 on Build GRCh38
rs751370659 Chr.14: 76166647-76166647 on Build GRCh38
rs749820660 Chr.14: 76166714-76166714 on Build GRCh38
rs751017163 Chr.14: 76166646-76166654 on Build GRCh38
rs757603055 Chr.14: 76166653-76166653 on Build GRCh38
rs985215736 Chr.14: 76166799-76166799 on Build GRCh38
rs780627676 Chr.14: 76166699-76166699 on Build GRCh38
rs777780130 Chr.14: 76166641-76166641 on Build GRCh38
rs751453563 Chr.14: 76166596-76166596 on Build GRCh38
rs762140090 Chr.14: 76166722-76166722 on Build GRCh38
rs569634138 Chr.14: 76166669-76166669 on Build GRCh38
rs368346751 Chr.14: 76166762-76166762 on Build GRCh38
rs146721749 Chr.14: 76166675-76166675 on Build GRCh38
rs758920954 Chr.14: 76166623-76166623 on Build GRCh38
rs920821133 Chr.14: 76166790-76166790 on Build GRCh38
rs752508096 Chr.14: 76166631-76166631 on Build GRCh38
rs374811109 Chr.14: 76166743-76166743 on Build GRCh38
rs371696590 Chr.14: 76166736-76166736 on Build GRCh38
rs767789682 Chr.14: 76166774-76166774 on Build GRCh38
rs770588601 Chr.14: 76166732-76166732 on Build GRCh38
rs971522052 Chr.14: 76166677-76166677 on Build GRCh38
rs955241952 Chr.14: 76166764-76166764 on Build GRCh38
rs745994861 Chr.14: 76166671-76166671 on Build GRCh38
rs1000091942 Chr.14: 76166588-76166588 on Build GRCh38
rs895304702 Chr.14: 76166586-76166586 on Build GRCh38
rs751422155 Chr.14: 76166761-76166761 on Build GRCh38
rs375395620 Chr.14: 76166696-76166696 on Build GRCh38
rs764102902 Chr.14: 76166756-76166756 on Build GRCh38
rs150810933 Chr.14: 76166637-76166637 on Build GRCh38
rs910946860 Chr.14: 76166802-76166802 on Build GRCh38
rs754857308 Chr.14: 76166621-76166621 on Build GRCh38
rs962470157 Chr.14: 76166804-76166804 on Build GRCh38

View Full Product Details