Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166365250-166365523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC1
Entrez Gene ID
51660
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L, CGI-129, MPYCD, dJ68L15.3
UniGene
Hs.172755
Gene Chromosome Location
Chr.6: 166364919-166383013 on Build GRCh38
Transcripts
NM_001270879.1 NM_016098.3 NR_073093.1 XM_011535894.1 XM_011535895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39878 c.319G>A p.A107T Chr.6: 166365440-166365440 on Build GRCh38
299079 c.306-8C>A p.? Chr.6: 166365461-166365461 on Build GRCh38
1184949 c.323C>A p.S108Y Chr.6: 166365436-166365436 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372437344 Chr.6: 166365395-166365395 on Build GRCh38
rs765354963 Chr.6: 166365471-166365471 on Build GRCh38
rs759728128 Chr.6: 166365453-166365453 on Build GRCh38
rs924863183 Chr.6: 166365330-166365330 on Build GRCh38
rs996088445 Chr.6: 166365366-166365366 on Build GRCh38
rs768193458 Chr.6: 166365398-166365398 on Build GRCh38
rs779377029 Chr.6: 166365390-166365390 on Build GRCh38
rs1029615916 Chr.6: 166365477-166365477 on Build GRCh38
rs866004713 Chr.6: 166365297-166365297 on Build GRCh38
rs189949541 Chr.6: 166365315-166365315 on Build GRCh38
rs762805358 Chr.6: 166365309-166365309 on Build GRCh38
rs934693237 Chr.6: 166365345-166365345 on Build GRCh38
rs139163077 Chr.6: 166365442-166365442 on Build GRCh38
rs775459967 Chr.6: 166365305-166365305 on Build GRCh38
rs748434808 Chr.6: 166365412-166365412 on Build GRCh38
rs146184945 Chr.6: 166365441-166365441 on Build GRCh38
rs773595678 Chr.6: 166365435-166365435 on Build GRCh38
rs774115517 Chr.6: 166365409-166365409 on Build GRCh38
rs748838366 Chr.6: 166365391-166365391 on Build GRCh38
rs763817746 Chr.6: 166365451-166365451 on Build GRCh38
rs772462474 Chr.6: 166365427-166365427 on Build GRCh38
rs773420004 Chr.6: 166365461-166365462 on Build GRCh38
rs1039953206 Chr.6: 166365432-166365432 on Build GRCh38
rs148611946 Chr.6: 166365375-166365375 on Build GRCh38
rs371125725 Chr.6: 166365389-166365389 on Build GRCh38
rs747267155 Chr.6: 166365486-166365489 on Build GRCh38
rs1010399226 Chr.6: 166365485-166365485 on Build GRCh38

View Full Product Details