Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 61499027-61499300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH20
Entrez Gene ID
28316
Gene Name
cadherin 20
Gene Aliases
CDH7L3, Cdh7
UniGene
Hs.671510
Gene Chromosome Location
Chr.18: 61333582-61555773 on Build GRCh38
Transcripts
NM_031891.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2881221 c.297G>A p.S99S Chr.18: 61499236-61499236 on Build GRCh38
4072831 c.262G>A p.D88N Chr.18: 61499201-61499201 on Build GRCh38
4543659 c.341G>A p.G114E Chr.18: 61499280-61499280 on Build GRCh38
2881220 c.271G>C p.D91H Chr.18: 61499210-61499210 on Build GRCh38
2881219 c.266G>A p.R89K Chr.18: 61499205-61499205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752079413 Chr.18: 61499242-61499242 on Build GRCh38
rs372146855 Chr.18: 61499060-61499060 on Build GRCh38
rs76457811 Chr.18: 61499155-61499155 on Build GRCh38
rs759745164 Chr.18: 61499227-61499227 on Build GRCh38
rs541180836 Chr.18: 61499269-61499269 on Build GRCh38
rs140950106 Chr.18: 61499266-61499266 on Build GRCh38
rs762246689 Chr.18: 61499246-61499246 on Build GRCh38
rs892172702 Chr.18: 61499134-61499134 on Build GRCh38
rs1019200569 Chr.18: 61499168-61499168 on Build GRCh38
rs967631515 Chr.18: 61499115-61499115 on Build GRCh38
rs755778264 Chr.18: 61499260-61499260 on Build GRCh38
rs750794751 Chr.18: 61499255-61499255 on Build GRCh38
rs770046815 Chr.18: 61499229-61499229 on Build GRCh38
rs372359014 Chr.18: 61499235-61499235 on Build GRCh38
rs150072548 Chr.18: 61499212-61499212 on Build GRCh38
rs145333204 Chr.18: 61499233-61499233 on Build GRCh38
rs151023560 Chr.18: 61499068-61499068 on Build GRCh38
rs1018865298 Chr.18: 61499083-61499083 on Build GRCh38
rs778962704 Chr.18: 61499146-61499146 on Build GRCh38
rs748050937 Chr.18: 61499147-61499147 on Build GRCh38
rs768524701 Chr.18: 61499140-61499140 on Build GRCh38
rs764114855 Chr.18: 61499236-61499236 on Build GRCh38
rs144277190 Chr.18: 61499270-61499270 on Build GRCh38
rs182572809 Chr.18: 61499073-61499073 on Build GRCh38
rs1022275952 Chr.18: 61499079-61499079 on Build GRCh38
rs377136301 Chr.18: 61499249-61499249 on Build GRCh38

View Full Product Details