Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4763208-4763437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT8
Entrez Gene ID
26290
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
GALNAC-T8
UniGene
Hs.511985
Gene Chromosome Location
Chr.12: 4720586-4772726 on Build GRCh38
Transcripts
NM_017417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
939815 c.1426A>C p.T476P Chr.12: 4763319-4763319 on Build GRCh38
3871706 c.1427C>T p.T476I Chr.12: 4763320-4763320 on Build GRCh38
6669131 c.1360-1G>T p.? Chr.12: 4763252-4763252 on Build GRCh38
1253050 c.1408G>T p.E470* Chr.12: 4763301-4763301 on Build GRCh38
3871704 c.1379G>A p.G460E Chr.12: 4763272-4763272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372477539 Chr.12: 4763298-4763298 on Build GRCh38
rs758180784 Chr.12: 4763333-4763333 on Build GRCh38
rs772532611 Chr.12: 4763398-4763398 on Build GRCh38
rs757359938 Chr.12: 4763307-4763307 on Build GRCh38
rs906982093 Chr.12: 4763389-4763389 on Build GRCh38
rs768244210 Chr.12: 4763343-4763343 on Build GRCh38
rs914455892 Chr.12: 4763317-4763317 on Build GRCh38
rs1035331646 Chr.12: 4763326-4763326 on Build GRCh38
rs36007275 Chr.12: 4763301-4763302 on Build GRCh38
rs760027164 Chr.12: 4763396-4763396 on Build GRCh38
rs768033819 Chr.12: 4763249-4763249 on Build GRCh38
rs780018727 Chr.12: 4763334-4763334 on Build GRCh38
rs34743826 Chr.12: 4763268-4763268 on Build GRCh38
rs1011779990 Chr.12: 4763277-4763277 on Build GRCh38
rs573950650 Chr.12: 4763380-4763380 on Build GRCh38
rs867147973 Chr.12: 4763402-4763402 on Build GRCh38
rs761333196 Chr.12: 4763260-4763260 on Build GRCh38
rs370583462 Chr.12: 4763316-4763316 on Build GRCh38
rs766873811 Chr.12: 4763262-4763262 on Build GRCh38
rs202099406 Chr.12: 4763252-4763252 on Build GRCh38
rs112230708 Chr.12: 4763251-4763251 on Build GRCh38
rs759973946 Chr.12: 4763286-4763286 on Build GRCh38
rs144204056 Chr.12: 4763291-4763291 on Build GRCh38
rs552155418 Chr.12: 4763386-4763386 on Build GRCh38
rs760565958 Chr.12: 4763337-4763337 on Build GRCh38
rs540716446 Chr.12: 4763239-4763239 on Build GRCh38
rs746592002 Chr.12: 4763248-4763248 on Build GRCh38
rs764036229 Chr.12: 4763304-4763304 on Build GRCh38
rs768552162 Chr.12: 4763405-4763406 on Build GRCh38
rs769635631 Chr.12: 4763392-4763392 on Build GRCh38
rs780520873 Chr.12: 4763375-4763375 on Build GRCh38
rs771788137 Chr.12: 4763267-4763267 on Build GRCh38
rs776091764 Chr.12: 4763400-4763400 on Build GRCh38
rs746889625 Chr.12: 4763335-4763335 on Build GRCh38
rs774800223 Chr.12: 4763276-4763276 on Build GRCh38
rs377514827 Chr.12: 4763306-4763306 on Build GRCh38
rs753233108 Chr.12: 4763297-4763297 on Build GRCh38
rs750288370 Chr.12: 4763328-4763328 on Build GRCh38
rs998454044 Chr.12: 4763321-4763321 on Build GRCh38
rs772984453 Chr.12: 4763394-4763394 on Build GRCh38
rs775430570 Chr.12: 4763284-4763284 on Build GRCh38

View Full Product Details