Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 345773-346004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP22
Entrez Gene ID
56940
Gene Name
dual specificity phosphatase 22
Gene Aliases
JKAP, JSP-1, JSP1, LMW-DSP2, LMWDSP2, MKP-x, MKPX, VHX
UniGene
Hs.29106
Gene Chromosome Location
Chr.6: 292057-351355 on Build GRCh38
Transcripts
NM_001286555.1 NM_020185.4 NR_104473.1 NR_104474.1 NR_104475.1 XM_011514757.2 XM_017011062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679718 c.52C>T p.R18C Chr.6: 345905-345905 on Build GRCh38
6650350 c.243T>C p.G81G Chr.6: 345908-345908 on Build GRCh38
5669193 c.226G>T p.E76* Chr.6: 345891-345891 on Build GRCh38
5693773 c.189-3C>G p.? Chr.6: 345851-345851 on Build GRCh38
3350941 c.238C>T p.R80C Chr.6: 345903-345903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749177335 Chr.6: 345867-345870 on Build GRCh38
rs1001711035 Chr.6: 345925-345925 on Build GRCh38
rs775466279 Chr.6: 345877-345877 on Build GRCh38
rs983908674 Chr.6: 345863-345863 on Build GRCh38
rs753528769 Chr.6: 345803-345803 on Build GRCh38
rs200764547 Chr.6: 345962-345962 on Build GRCh38
rs764036547 Chr.6: 345850-345850 on Build GRCh38
rs761572900 Chr.6: 345806-345806 on Build GRCh38
rs751615989 Chr.6: 345969-345969 on Build GRCh38
rs754390377 Chr.6: 345954-345954 on Build GRCh38
rs529856867 Chr.6: 345826-345826 on Build GRCh38
rs140808686 Chr.6: 345904-345904 on Build GRCh38
rs780002043 Chr.6: 345836-345837 on Build GRCh38
rs765852853 Chr.6: 345935-345935 on Build GRCh38
rs374233441 Chr.6: 345897-345897 on Build GRCh38
rs369153482 Chr.6: 345890-345890 on Build GRCh38
rs1025631164 Chr.6: 345940-345940 on Build GRCh38
rs778054165 Chr.6: 345839-345839 on Build GRCh38
rs746898656 Chr.6: 345919-345919 on Build GRCh38
rs756588736 Chr.6: 345899-345899 on Build GRCh38
rs773051965 Chr.6: 345894-345894 on Build GRCh38
rs755496719 Chr.6: 345824-345824 on Build GRCh38
rs376941361 Chr.6: 345933-345933 on Build GRCh38
rs370568995 Chr.6: 345936-345936 on Build GRCh38
rs746977383 Chr.6: 345880-345880 on Build GRCh38
rs560306784 Chr.6: 345827-345827 on Build GRCh38
rs956883288 Chr.6: 345939-345939 on Build GRCh38
rs566311245 Chr.6: 345849-345849 on Build GRCh38
rs764775470 Chr.6: 345808-345808 on Build GRCh38
rs756089358 Chr.6: 345837-345841 on Build GRCh38
rs776521161 Chr.6: 345926-345926 on Build GRCh38
rs745968465 Chr.6: 345856-345856 on Build GRCh38
rs749450706 Chr.6: 345844-345844 on Build GRCh38
rs909743050 Chr.6: 345875-345875 on Build GRCh38
rs761519812 Chr.6: 345891-345891 on Build GRCh38
rs764372378 Chr.6: 345900-345900 on Build GRCh38
rs145392896 Chr.6: 345893-345893 on Build GRCh38
rs200834679 Chr.6: 345903-345903 on Build GRCh38
rs137855591 Chr.6: 345896-345896 on Build GRCh38
rs368122798 Chr.6: 345953-345953 on Build GRCh38
rs756492316 Chr.6: 345838-345838 on Build GRCh38
rs761202005 Chr.6: 345944-345944 on Build GRCh38
rs144845390 Chr.6: 345905-345905 on Build GRCh38
rs148279164 Chr.6: 345906-345906 on Build GRCh38
rs373954135 Chr.6: 345932-345932 on Build GRCh38
rs3823302 Chr.6: 345911-345911 on Build GRCh38
rs199871079 Chr.6: 345972-345972 on Build GRCh38
rs377115548 Chr.6: 345898-345898 on Build GRCh38
rs201410036 Chr.6: 345973-345973 on Build GRCh38
rs755137431 Chr.6: 345970-345970 on Build GRCh38
rs755066347 Chr.6: 345930-345931 on Build GRCh38
rs749986738 Chr.6: 345822-345822 on Build GRCh38
rs768442120 Chr.6: 345884-345884 on Build GRCh38
rs918649739 Chr.6: 345829-345829 on Build GRCh38
rs368343297 Chr.6: 345968-345968 on Build GRCh38
rs768508855 Chr.6: 345920-345920 on Build GRCh38
rs780011974 Chr.6: 345916-345916 on Build GRCh38
rs772091549 Chr.6: 345874-345874 on Build GRCh38

View Full Product Details