Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32078969-32079208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRY
Entrez Gene ID
10129
Gene Name
FRY microtubule binding protein
Gene Aliases
13CDNA73, 214K23.2, C13orf14, CG003, bA207N4.2, bA37E23.1
UniGene
Hs.507669
Gene Chromosome Location
Chr.13: 32029742-32299122 on Build GRCh38
Transcripts
NM_023037.2 XM_006719749.3 XM_017020303.1 XM_017020304.1 XM_017020305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6074252 c.256C>T p.P86S Chr.13: 32079028-32079028 on Build GRCh38
3468322 c.247C>T p.R83C Chr.13: 32079010-32079010 on Build GRCh38
6667315 c.239G>A p.R80H Chr.13: 32079011-32079011 on Build GRCh38
4641813 c.201A>G p.L67L Chr.13: 32078973-32078973 on Build GRCh38
5952839 c.247C>G p.R83G Chr.13: 32079010-32079010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141122444 Chr.13: 32079061-32079062 on Build GRCh38
rs76615559 Chr.13: 32079105-32079105 on Build GRCh38
rs75678839 Chr.13: 32079166-32079166 on Build GRCh38
rs759496780 Chr.13: 32079001-32079001 on Build GRCh38
rs796748766 Chr.13: 32079062-32079063 on Build GRCh38
rs984334491 Chr.13: 32079023-32079023 on Build GRCh38
rs55977206 Chr.13: 32079106-32079106 on Build GRCh38
rs55830995 Chr.13: 32079160-32079160 on Build GRCh38
rs942449623 Chr.13: 32079100-32079100 on Build GRCh38
rs376716820 Chr.13: 32079008-32079008 on Build GRCh38
rs1026466773 Chr.13: 32079027-32079027 on Build GRCh38
rs201690345 Chr.13: 32079062-32079062 on Build GRCh38
rs758211187 Chr.13: 32079030-32079030 on Build GRCh38
rs796464207 Chr.13: 32079061-32079062 on Build GRCh38
rs978135267 Chr.13: 32079154-32079154 on Build GRCh38
rs910972192 Chr.13: 32079073-32079073 on Build GRCh38
rs368105613 Chr.13: 32079063-32079064 on Build GRCh38
rs559547593 Chr.13: 32079063-32079063 on Build GRCh38
rs894603058 Chr.13: 32079177-32079177 on Build GRCh38
rs573441706 Chr.13: 32079036-32079036 on Build GRCh38
rs745323573 Chr.13: 32079055-32079055 on Build GRCh38
rs946314416 Chr.13: 32079114-32079114 on Build GRCh38
rs906303042 Chr.13: 32079039-32079039 on Build GRCh38
rs904743515 Chr.13: 32079146-32079146 on Build GRCh38
rs752479049 Chr.13: 32079022-32079022 on Build GRCh38
rs79025279 Chr.13: 32079144-32079144 on Build GRCh38
rs756904665 Chr.13: 32079044-32079044 on Build GRCh38
rs545616571 Chr.13: 32079047-32079047 on Build GRCh38

View Full Product Details