Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64989299-64989572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BATF2
Entrez Gene ID
116071
Gene Name
basic leucine zipper ATF-like transcription factor 2
Gene Aliases
SARI
UniGene
Hs.124840
Gene Chromosome Location
Chr.11: 64987945-64997045 on Build GRCh38
Transcripts
NM_001300807.1 NM_001300808.1 NM_138456.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451662 c.559G>A p.G187R Chr.11: 64989395-64989395 on Build GRCh38
4647408 c.553C>A p.L185M Chr.11: 64989401-64989401 on Build GRCh38
1245990 c.638_639insGCA p.E212_H213insQ Chr.11: 64989315-64989316 on Build GRCh38
1355856 c.582C>A p.A194A Chr.11: 64989372-64989372 on Build GRCh38
6134070 c.487G>A p.V163I Chr.11: 64989467-64989467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772143507 Chr.11: 64989518-64989518 on Build GRCh38
rs749049969 Chr.11: 64989414-64989414 on Build GRCh38
rs777881057 Chr.11: 64989399-64989399 on Build GRCh38
rs148209044 Chr.11: 64989540-64989540 on Build GRCh38
rs370857140 Chr.11: 64989373-64989373 on Build GRCh38
rs755458467 Chr.11: 64989370-64989370 on Build GRCh38
rs146768788 Chr.11: 64989340-64989340 on Build GRCh38
rs770006168 Chr.11: 64989546-64989546 on Build GRCh38
rs765650570 Chr.11: 64989381-64989381 on Build GRCh38
rs200380956 Chr.11: 64989398-64989398 on Build GRCh38
rs781375669 Chr.11: 64989482-64989482 on Build GRCh38
rs143080238 Chr.11: 64989534-64989534 on Build GRCh38
rs772964449 Chr.11: 64989343-64989343 on Build GRCh38
rs376983784 Chr.11: 64989473-64989473 on Build GRCh38
rs149025465 Chr.11: 64989505-64989505 on Build GRCh38
rs778815144 Chr.11: 64989512-64989512 on Build GRCh38
rs771683673 Chr.11: 64989431-64989431 on Build GRCh38
rs140399350 Chr.11: 64989354-64989354 on Build GRCh38
rs551804003 Chr.11: 64989421-64989421 on Build GRCh38
rs754263193 Chr.11: 64989355-64989355 on Build GRCh38
rs912451016 Chr.11: 64989492-64989492 on Build GRCh38
rs765674781 Chr.11: 64989470-64989470 on Build GRCh38
rs746509534 Chr.11: 64989532-64989532 on Build GRCh38
rs763279518 Chr.11: 64989515-64989516 on Build GRCh38
rs775870040 Chr.11: 64989486-64989494 on Build GRCh38
rs751729818 Chr.11: 64989407-64989407 on Build GRCh38
rs143335032 Chr.11: 64989327-64989327 on Build GRCh38
rs763537147 Chr.11: 64989474-64989474 on Build GRCh38
rs750721303 Chr.11: 64989493-64989493 on Build GRCh38
rs529390417 Chr.11: 64989341-64989341 on Build GRCh38
rs368995882 Chr.11: 64989418-64989418 on Build GRCh38
rs140401210 Chr.11: 64989348-64989348 on Build GRCh38
rs747999045 Chr.11: 64989513-64989513 on Build GRCh38
rs753399253 Chr.11: 64989458-64989458 on Build GRCh38
rs947429631 Chr.11: 64989545-64989545 on Build GRCh38
rs775283756 Chr.11: 64989326-64989326 on Build GRCh38
rs962380112 Chr.11: 64989378-64989378 on Build GRCh38
rs768550449 Chr.11: 64989417-64989417 on Build GRCh38
rs976495999 Chr.11: 64989498-64989498 on Build GRCh38
rs760165042 Chr.11: 64989339-64989339 on Build GRCh38
rs946970455 Chr.11: 64989542-64989542 on Build GRCh38
rs34403604 Chr.11: 64989329-64989329 on Build GRCh38
rs776116800 Chr.11: 64989536-64989536 on Build GRCh38
rs374430545 Chr.11: 64989440-64989440 on Build GRCh38
rs145280751 Chr.11: 64989444-64989444 on Build GRCh38
rs751641909 Chr.11: 64989478-64989478 on Build GRCh38
rs779757104 Chr.11: 64989413-64989413 on Build GRCh38
rs1036337917 Chr.11: 64989344-64989344 on Build GRCh38
rs537808630 Chr.11: 64989410-64989410 on Build GRCh38
rs770724887 Chr.11: 64989452-64989452 on Build GRCh38
rs1042185097 Chr.11: 64989550-64989550 on Build GRCh38
rs144900937 Chr.11: 64989451-64989451 on Build GRCh38
rs753313832 Chr.11: 64989397-64989397 on Build GRCh38
rs764118566 Chr.11: 64989477-64989477 on Build GRCh38
rs139572037 Chr.11: 64989320-64989320 on Build GRCh38

View Full Product Details