Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7096505-7096778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1RL
Entrez Gene ID
51279
Gene Name
complement C1r subcomponent like
Gene Aliases
C1RL1, C1RLP, C1r-LP, CLSPa
UniGene
Hs.631730
Gene Chromosome Location
Chr.12: 7093662-7109278 on Build GRCh38
Transcripts
NM_001297640.1 NM_001297642.1 NM_016546.3 XM_017019397.1 XM_017019398.1 XM_017019399.1 XM_017019400.1 XM_017019402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142889 c.1249G>T p.D417Y Chr.12: 7096606-7096606 on Build GRCh38
3465079 c.1146G>A p.M382I Chr.12: 7096709-7096709 on Build GRCh38
4854171 c.1141G>C p.E381Q Chr.12: 7096714-7096714 on Build GRCh38
159557 c.1122C>A p.Y374* Chr.12: 7096733-7096733 on Build GRCh38
4909851 c.1343A>G p.H448R Chr.12: 7096512-7096512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762191344 Chr.12: 7096707-7096707 on Build GRCh38
rs76391175 Chr.12: 7096547-7096547 on Build GRCh38
rs78865688 Chr.12: 7096548-7096548 on Build GRCh38
rs764528857 Chr.12: 7096606-7096606 on Build GRCh38
rs1029829287 Chr.12: 7096740-7096740 on Build GRCh38
rs759003415 Chr.12: 7096749-7096749 on Build GRCh38
rs773674388 Chr.12: 7096688-7096688 on Build GRCh38
rs764572014 Chr.12: 7096697-7096697 on Build GRCh38
rs769688821 Chr.12: 7096670-7096670 on Build GRCh38
rs867787655 Chr.12: 7096710-7096710 on Build GRCh38
rs759069993 Chr.12: 7096690-7096690 on Build GRCh38
rs777643209 Chr.12: 7096739-7096739 on Build GRCh38
rs772105163 Chr.12: 7096560-7096560 on Build GRCh38
rs964518700 Chr.12: 7096693-7096693 on Build GRCh38
rs765988783 Chr.12: 7096680-7096680 on Build GRCh38
rs192526191 Chr.12: 7096541-7096541 on Build GRCh38
rs772978274 Chr.12: 7096587-7096587 on Build GRCh38
rs769740883 Chr.12: 7096736-7096736 on Build GRCh38
rs753114917 Chr.12: 7096536-7096536 on Build GRCh38
rs142793242 Chr.12: 7096577-7096577 on Build GRCh38
rs150498892 Chr.12: 7096732-7096732 on Build GRCh38
rs1000037896 Chr.12: 7096700-7096700 on Build GRCh38
rs765549175 Chr.12: 7096708-7096708 on Build GRCh38
rs758559770 Chr.12: 7096714-7096714 on Build GRCh38
rs946065897 Chr.12: 7096616-7096616 on Build GRCh38
rs750651226 Chr.12: 7096712-7096712 on Build GRCh38
rs754209479 Chr.12: 7096613-7096613 on Build GRCh38
rs756508928 Chr.12: 7096604-7096604 on Build GRCh38
rs746869190 Chr.12: 7096571-7096571 on Build GRCh38
rs369232515 Chr.12: 7096645-7096645 on Build GRCh38
rs868407640 Chr.12: 7096733-7096733 on Build GRCh38
rs139459588 Chr.12: 7096753-7096753 on Build GRCh38
rs771340998 Chr.12: 7096578-7096578 on Build GRCh38
rs988504464 Chr.12: 7096552-7096552 on Build GRCh38
rs771566519 Chr.12: 7096752-7096752 on Build GRCh38
rs74323236 Chr.12: 7096590-7096590 on Build GRCh38
rs199891851 Chr.12: 7096646-7096646 on Build GRCh38
rs373433289 Chr.12: 7096675-7096675 on Build GRCh38
rs373012039 Chr.12: 7096619-7096619 on Build GRCh38
rs138873491 Chr.12: 7096540-7096540 on Build GRCh38
rs770544200 Chr.12: 7096747-7096747 on Build GRCh38
rs751104076 Chr.12: 7096704-7096704 on Build GRCh38
rs775434872 Chr.12: 7096570-7096570 on Build GRCh38
rs753276800 Chr.12: 7096601-7096601 on Build GRCh38
rs755055442 Chr.12: 7096719-7096719 on Build GRCh38
rs369851400 Chr.12: 7096618-7096618 on Build GRCh38
rs747995939 Chr.12: 7096663-7096663 on Build GRCh38
rs916949168 Chr.12: 7096553-7096553 on Build GRCh38
rs141631026 Chr.12: 7096718-7096718 on Build GRCh38
rs376313967 Chr.12: 7096629-7096629 on Build GRCh38
rs137984832 Chr.12: 7096716-7096716 on Build GRCh38
rs1029655149 Chr.12: 7096637-7096637 on Build GRCh38
rs774044185 Chr.12: 7096748-7096748 on Build GRCh38
rs765040924 Chr.12: 7096585-7096585 on Build GRCh38
rs77140636 Chr.12: 7096549-7096549 on Build GRCh38
rs949802369 Chr.12: 7096566-7096566 on Build GRCh38
rs748960287 Chr.12: 7096742-7096742 on Build GRCh38
rs765937376 Chr.12: 7096591-7096591 on Build GRCh38
rs779106623 Chr.12: 7096550-7096550 on Build GRCh38
rs779934100 Chr.12: 7096626-7096626 on Build GRCh38
rs758506687 Chr.12: 7096622-7096622 on Build GRCh38

View Full Product Details