Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101531194-101531419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTRC
Entrez Gene ID
8945
Gene Name
beta-transducin repeat containing E3 ubiquitin protein ligase
Gene Aliases
BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP
UniGene
Hs.643802
Gene Chromosome Location
Chr.10: 101353808-101557321 on Build GRCh38
Transcripts
NM_001256856.1 NM_003939.4 NM_033637.3 XM_006718054.2 XM_011540320.2 XM_017016870.1 XM_017016871.1 XM_017016872.1 XM_017016873.1 XM_017016874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419768 c.809C>A p.A270E Chr.10: 101531302-101531302 on Build GRCh38
6295470 c.780G>T p.G260G Chr.10: 101531273-101531273 on Build GRCh38
6613631 c.744-8delT p.? Chr.10: 101531229-101531229 on Build GRCh38
4666416 c.833A>G p.D278G Chr.10: 101531326-101531326 on Build GRCh38
5991331 c.836T>C p.I279T Chr.10: 101531329-101531329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760847771 Chr.10: 101531267-101531267 on Build GRCh38
rs765768695 Chr.10: 101531300-101531300 on Build GRCh38
rs371692654 Chr.10: 101531270-101531270 on Build GRCh38
rs758785153 Chr.10: 101531271-101531271 on Build GRCh38
rs146435684 Chr.10: 101531321-101531321 on Build GRCh38
rs1003064016 Chr.10: 101531358-101531358 on Build GRCh38
rs201756663 Chr.10: 101531239-101531239 on Build GRCh38
rs886865434 Chr.10: 101531382-101531382 on Build GRCh38
rs899386918 Chr.10: 101531322-101531322 on Build GRCh38
rs188592989 Chr.10: 101531354-101531354 on Build GRCh38
rs1011810382 Chr.10: 101531310-101531310 on Build GRCh38
rs755177621 Chr.10: 101531361-101531362 on Build GRCh38
rs755774420 Chr.10: 101531362-101531362 on Build GRCh38
rs771355300 Chr.10: 101531393-101531393 on Build GRCh38
rs779126209 Chr.10: 101531371-101531371 on Build GRCh38
rs556352720 Chr.10: 101531344-101531344 on Build GRCh38
rs367662470 Chr.10: 101531245-101531245 on Build GRCh38
rs756766939 Chr.10: 101531323-101531323 on Build GRCh38
rs1023233452 Chr.10: 101531342-101531342 on Build GRCh38
rs767683874 Chr.10: 101531249-101531249 on Build GRCh38
rs751086036 Chr.10: 101531304-101531304 on Build GRCh38
rs780004511 Chr.10: 101531369-101531369 on Build GRCh38
rs773452364 Chr.10: 101531251-101531251 on Build GRCh38
rs754367468 Chr.10: 101531291-101531292 on Build GRCh38
rs756177557 Chr.10: 101531341-101531342 on Build GRCh38
rs1007968813 Chr.10: 101531391-101531391 on Build GRCh38
rs755477503 Chr.10: 101531286-101531286 on Build GRCh38

View Full Product Details