Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50818154-50818381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS4
Entrez Gene ID
55653
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
CNOL
UniGene
Hs.381178
Gene Chromosome Location
Chr.20: 50794894-50877736 on Build GRCh38
Transcripts
NM_001010974.2 NM_017843.4 NM_198799.3 XM_011528886.2 XM_011528887.2 XM_017027932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7208856 c.202A>G p.I68V Chr.20: 50818232-50818232 on Build GRCh38
5430031 c.229G>C p.E77Q Chr.20: 50818259-50818259 on Build GRCh38
4473126 c.182C>G p.A61G Chr.20: 50818212-50818212 on Build GRCh38
3547653 c.183G>A p.A61A Chr.20: 50818213-50818213 on Build GRCh38
3547656 c.192G>A p.V64V Chr.20: 50818222-50818222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766472880 Chr.20: 50818230-50818230 on Build GRCh38
rs776911082 Chr.20: 50818212-50818212 on Build GRCh38
rs756482629 Chr.20: 50818302-50818303 on Build GRCh38
rs988884266 Chr.20: 50818224-50818224 on Build GRCh38
rs776344299 Chr.20: 50818307-50818307 on Build GRCh38
rs1057021465 Chr.20: 50818352-50818352 on Build GRCh38
rs758848754 Chr.20: 50818225-50818225 on Build GRCh38
rs190664502 Chr.20: 50818246-50818246 on Build GRCh38
rs539355212 Chr.20: 50818198-50818198 on Build GRCh38
rs775090668 Chr.20: 50818197-50818197 on Build GRCh38
rs779749118 Chr.20: 50818282-50818282 on Build GRCh38
rs965376843 Chr.20: 50818222-50818222 on Build GRCh38
rs772094503 Chr.20: 50818278-50818278 on Build GRCh38
rs995673229 Chr.20: 50818177-50818177 on Build GRCh38
rs759655085 Chr.20: 50818324-50818324 on Build GRCh38
rs915723513 Chr.20: 50818239-50818239 on Build GRCh38
rs745821809 Chr.20: 50818255-50818255 on Build GRCh38
rs201012008 Chr.20: 50818326-50818326 on Build GRCh38
rs11283878 Chr.20: 50818292-50818293 on Build GRCh38
rs773520407 Chr.20: 50818317-50818317 on Build GRCh38
rs752974212 Chr.20: 50818333-50818333 on Build GRCh38
rs751600956 Chr.20: 50818231-50818231 on Build GRCh38
rs780451801 Chr.20: 50818314-50818315 on Build GRCh38
rs763485122 Chr.20: 50818321-50818321 on Build GRCh38
rs201932513 Chr.20: 50818234-50818234 on Build GRCh38
rs768252686 Chr.20: 50818303-50818303 on Build GRCh38
rs750669757 Chr.20: 50818291-50818291 on Build GRCh38
rs770209249 Chr.20: 50818316-50818316 on Build GRCh38
rs745729982 Chr.20: 50818181-50818181 on Build GRCh38
rs766844311 Chr.20: 50818322-50818322 on Build GRCh38
rs778882207 Chr.20: 50818317-50818318 on Build GRCh38
rs377675495 Chr.20: 50818213-50818213 on Build GRCh38
rs570332652 Chr.20: 50818195-50818195 on Build GRCh38
rs370907861 Chr.20: 50818235-50818235 on Build GRCh38
rs746493565 Chr.20: 50818301-50818301 on Build GRCh38
rs899202056 Chr.20: 50818353-50818353 on Build GRCh38
rs751692923 Chr.20: 50818323-50818323 on Build GRCh38
rs761304944 Chr.20: 50818313-50818313 on Build GRCh38
rs376032771 Chr.20: 50818316-50818317 on Build GRCh38
rs148903431 Chr.20: 50818242-50818242 on Build GRCh38
rs200095102 Chr.20: 50818249-50818249 on Build GRCh38
rs532303612 Chr.20: 50818241-50818241 on Build GRCh38

View Full Product Details