Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171522497-171522750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBRD1
Entrez Gene ID
79901
Gene Name
cytochrome b reductase 1
Gene Aliases
CYB561A2, DCYTB, FRRS3
UniGene
Hs.221941
Gene Chromosome Location
Chr.2: 171522247-171558133 on Build GRCh38
Transcripts
NM_001127383.1 NM_001256909.1 NM_024843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3569989 c.180C>T p.F60F Chr.2: 171522725-171522725 on Build GRCh38
6412591 c.169G>A p.G57S Chr.2: 171522714-171522714 on Build GRCh38
3714038 c.89G>T p.W30L Chr.2: 171522634-171522634 on Build GRCh38
5690485 c.136G>C p.E46Q Chr.2: 171522681-171522681 on Build GRCh38
4470917 c.168C>G p.T56T Chr.2: 171522713-171522713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775711731 Chr.2: 171522649-171522649 on Build GRCh38
rs756288449 Chr.2: 171522611-171522611 on Build GRCh38
rs1017584530 Chr.2: 171522678-171522678 on Build GRCh38
rs372271042 Chr.2: 171522587-171522587 on Build GRCh38
rs564331627 Chr.2: 171522598-171522598 on Build GRCh38
rs774947139 Chr.2: 171522567-171522567 on Build GRCh38
rs746335457 Chr.2: 171522640-171522640 on Build GRCh38
rs750321364 Chr.2: 171522591-171522591 on Build GRCh38
rs779171072 Chr.2: 171522630-171522630 on Build GRCh38
rs139365788 Chr.2: 171522572-171522572 on Build GRCh38
rs752248111 Chr.2: 171522518-171522518 on Build GRCh38
rs776216750 Chr.2: 171522674-171522674 on Build GRCh38
rs201951213 Chr.2: 171522543-171522543 on Build GRCh38
rs769020511 Chr.2: 171522667-171522667 on Build GRCh38
rs772289853 Chr.2: 171522577-171522577 on Build GRCh38
rs753402712 Chr.2: 171522531-171522531 on Build GRCh38
rs761485444 Chr.2: 171522677-171522677 on Build GRCh38
rs868480546 Chr.2: 171522726-171522726 on Build GRCh38
rs757181239 Chr.2: 171522535-171522535 on Build GRCh38
rs758079844 Chr.2: 171522549-171522549 on Build GRCh38
rs771143937 Chr.2: 171522563-171522563 on Build GRCh38
rs763486773 Chr.2: 171522524-171522524 on Build GRCh38
rs199621126 Chr.2: 171522596-171522596 on Build GRCh38
rs761032280 Chr.2: 171522656-171522656 on Build GRCh38
rs764696712 Chr.2: 171522692-171522692 on Build GRCh38
rs778755032 Chr.2: 171522542-171522542 on Build GRCh38
rs775970725 Chr.2: 171522579-171522579 on Build GRCh38
rs149428156 Chr.2: 171522602-171522602 on Build GRCh38
rs201423832 Chr.2: 171522552-171522552 on Build GRCh38
rs146713767 Chr.2: 171522589-171522589 on Build GRCh38
rs565938208 Chr.2: 171522590-171522590 on Build GRCh38
rs771090771 Chr.2: 171522629-171522629 on Build GRCh38
rs754748261 Chr.2: 171522720-171522720 on Build GRCh38
rs767411280 Chr.2: 171522614-171522614 on Build GRCh38
rs767447864 Chr.2: 171522723-171522723 on Build GRCh38
rs553503102 Chr.2: 171522707-171522707 on Build GRCh38
rs1014505885 Chr.2: 171522544-171522544 on Build GRCh38
rs779693212 Chr.2: 171522600-171522600 on Build GRCh38
rs749449959 Chr.2: 171522561-171522561 on Build GRCh38
rs201989257 Chr.2: 171522648-171522648 on Build GRCh38
rs200632435 Chr.2: 171522714-171522714 on Build GRCh38
rs770105209 Chr.2: 171522558-171522558 on Build GRCh38
rs766408302 Chr.2: 171522711-171522711 on Build GRCh38
rs755828772 Chr.2: 171522521-171522521 on Build GRCh38
rs777988908 Chr.2: 171522560-171522560 on Build GRCh38
rs749538088 Chr.2: 171522626-171522626 on Build GRCh38
rs749999527 Chr.2: 171522694-171522694 on Build GRCh38
rs991177451 Chr.2: 171522689-171522689 on Build GRCh38
rs762788179 Chr.2: 171522710-171522710 on Build GRCh38

View Full Product Details