Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74135508-74135755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLA3
Entrez Gene ID
388962
Gene Name
bolA family member 3
Gene Aliases
MMDS2
UniGene
Hs.61472
Gene Chromosome Location
Chr.2: 74135401-74147912 on Build GRCh38
Transcripts
NM_001035505.1 NM_212552.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343703 c.271C>A p.P91T Chr.2: 74135557-74135557 on Build GRCh38
1022911 c.313C>T p.P105S Chr.2: 74135604-74135604 on Build GRCh38
6612269 c.259-1G>T p.? Chr.2: 74135659-74135659 on Build GRCh38
1022910 c.314C>T p.P105L Chr.2: 74135603-74135603 on Build GRCh38
4874558 c.224C>T p.P75L Chr.2: 74135604-74135604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766621227 Chr.2: 74135558-74135558 on Build GRCh38
rs148674363 Chr.2: 74135630-74135630 on Build GRCh38
rs751495007 Chr.2: 74135691-74135691 on Build GRCh38
rs773696418 Chr.2: 74135649-74135649 on Build GRCh38
rs766637713 Chr.2: 74135662-74135662 on Build GRCh38
rs759490061 Chr.2: 74135675-74135675 on Build GRCh38
rs146584766 Chr.2: 74135720-74135720 on Build GRCh38
rs769219293 Chr.2: 74135620-74135620 on Build GRCh38
rs142868057 Chr.2: 74135676-74135676 on Build GRCh38
rs908781045 Chr.2: 74135628-74135628 on Build GRCh38
rs150626228 Chr.2: 74135598-74135598 on Build GRCh38
rs549931538 Chr.2: 74135624-74135624 on Build GRCh38
rs923996415 Chr.2: 74135693-74135693 on Build GRCh38
rs191936501 Chr.2: 74135589-74135589 on Build GRCh38
rs751396525 Chr.2: 74135585-74135585 on Build GRCh38
rs376876176 Chr.2: 74135600-74135600 on Build GRCh38
rs750269271 Chr.2: 74135588-74135588 on Build GRCh38
rs753960028 Chr.2: 74135565-74135565 on Build GRCh38
rs146336257 Chr.2: 74135597-74135597 on Build GRCh38
rs566060433 Chr.2: 74135699-74135699 on Build GRCh38
rs886056326 Chr.2: 74135562-74135562 on Build GRCh38
rs941523861 Chr.2: 74135642-74135642 on Build GRCh38
rs777509576 Chr.2: 74135580-74135580 on Build GRCh38
rs202046520 Chr.2: 74135621-74135621 on Build GRCh38
rs756966193 Chr.2: 74135586-74135586 on Build GRCh38
rs758410967 Chr.2: 74135572-74135572 on Build GRCh38
rs866949521 Chr.2: 74135622-74135622 on Build GRCh38
rs200938488 Chr.2: 74135635-74135635 on Build GRCh38
rs761041660 Chr.2: 74135651-74135651 on Build GRCh38
rs776860491 Chr.2: 74135674-74135674 on Build GRCh38

View Full Product Details