Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73869937-73870162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL1
Entrez Gene ID
2919
Gene Name
C-X-C motif chemokine ligand 1
Gene Aliases
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
UniGene
Hs.708652
Gene Chromosome Location
Chr.4: 73869392-73871302 on Build GRCh38
Transcripts
NM_001511.3 NR_046035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201302 c.292G>A p.E98K Chr.4: 73869973-73869973 on Build GRCh38
3428649 c.293A>G p.E98G Chr.4: 73869974-73869974 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372229384 Chr.4: 73870018-73870018 on Build GRCh38
rs762980706 Chr.4: 73869985-73869985 on Build GRCh38
rs922536633 Chr.4: 73870047-73870047 on Build GRCh38
rs777740404 Chr.4: 73870043-73870043 on Build GRCh38
rs2018732 Chr.4: 73870017-73870017 on Build GRCh38
rs181532013 Chr.4: 73870098-73870098 on Build GRCh38
rs199517728 Chr.4: 73869988-73869988 on Build GRCh38
rs201976269 Chr.4: 73870034-73870034 on Build GRCh38
rs549189473 Chr.4: 73869987-73869987 on Build GRCh38
rs1034458362 Chr.4: 73870093-73870093 on Build GRCh38
rs201003859 Chr.4: 73869973-73869973 on Build GRCh38
rs755248483 Chr.4: 73870001-73870001 on Build GRCh38
rs756177188 Chr.4: 73870035-73870035 on Build GRCh38
rs200517725 Chr.4: 73869998-73869998 on Build GRCh38
rs759232302 Chr.4: 73869972-73869972 on Build GRCh38
rs201361001 Chr.4: 73869979-73869979 on Build GRCh38
rs200131602 Chr.4: 73870028-73870028 on Build GRCh38
rs752313297 Chr.4: 73869975-73869975 on Build GRCh38
rs993006785 Chr.4: 73870108-73870108 on Build GRCh38
rs371670459 Chr.4: 73869996-73869996 on Build GRCh38
rs778955268 Chr.4: 73870007-73870007 on Build GRCh38
rs528161039 Chr.4: 73870077-73870077 on Build GRCh38
rs1024553191 Chr.4: 73870123-73870123 on Build GRCh38
rs748264294 Chr.4: 73870010-73870010 on Build GRCh38
rs756956042 Chr.4: 73869994-73869994 on Build GRCh38
rs571152914 Chr.4: 73870099-73870099 on Build GRCh38
rs538633260 Chr.4: 73870130-73870130 on Build GRCh38
rs1049390051 Chr.4: 73870140-73870140 on Build GRCh38
rs200649425 Chr.4: 73870031-73870031 on Build GRCh38

View Full Product Details