Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97307619-97307846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC180
Entrez Gene ID
100499483
Gene Name
coiled-coil domain containing 180
Gene Aliases
BDAG1, C9orf174, KIAA1529
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97307628-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5201500 c.406C>T p.R136W Chr.9: 97307744-97307744 on Build GRCh38
245256 c.429G>A p.G143G Chr.9: 97307767-97307767 on Build GRCh38
5063423 c.7G>A p.G3S Chr.9: 97307762-97307762 on Build GRCh38
6621947 c.422G>A p.R141H Chr.9: 97307760-97307760 on Build GRCh38
6621949 c.422G>A p.R141H Chr.9: 97307760-97307760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752389077 Chr.9: 97307688-97307689 on Build GRCh38
rs76020873 Chr.9: 97307664-97307664 on Build GRCh38
rs757116613 Chr.9: 97307776-97307776 on Build GRCh38
rs937958533 Chr.9: 97307643-97307643 on Build GRCh38
rs201031305 Chr.9: 97307784-97307784 on Build GRCh38
rs749003415 Chr.9: 97307775-97307775 on Build GRCh38
rs773641972 Chr.9: 97307695-97307695 on Build GRCh38
rs193093838 Chr.9: 97307686-97307686 on Build GRCh38
rs900896042 Chr.9: 97307732-97307732 on Build GRCh38
rs778579527 Chr.9: 97307778-97307778 on Build GRCh38
rs563429380 Chr.9: 97307773-97307773 on Build GRCh38
rs796074467 Chr.9: 97307746-97307746 on Build GRCh38
rs752036247 Chr.9: 97307759-97307759 on Build GRCh38
rs974207267 Chr.9: 97307702-97307702 on Build GRCh38
rs375088978 Chr.9: 97307714-97307714 on Build GRCh38
rs766789247 Chr.9: 97307758-97307758 on Build GRCh38
rs763164355 Chr.9: 97307801-97307801 on Build GRCh38
rs749056531 Chr.9: 97307706-97307706 on Build GRCh38
rs755721446 Chr.9: 97307765-97307765 on Build GRCh38
rs148245431 Chr.9: 97307788-97307788 on Build GRCh38
rs917878561 Chr.9: 97307757-97307757 on Build GRCh38
rs765475594 Chr.9: 97307753-97307753 on Build GRCh38
rs143152642 Chr.9: 97307756-97307756 on Build GRCh38
rs748196889 Chr.9: 97307787-97307787 on Build GRCh38
rs773411856 Chr.9: 97307799-97307799 on Build GRCh38
rs145118939 Chr.9: 97307763-97307763 on Build GRCh38
rs774329532 Chr.9: 97307723-97307723 on Build GRCh38
rs985780570 Chr.9: 97307650-97307650 on Build GRCh38
rs767002900 Chr.9: 97307762-97307762 on Build GRCh38
rs755852787 Chr.9: 97307691-97307691 on Build GRCh38
rs140624448 Chr.9: 97307781-97307781 on Build GRCh38
rs577188983 Chr.9: 97307744-97307744 on Build GRCh38
rs768718102 Chr.9: 97307740-97307740 on Build GRCh38
rs745863621 Chr.9: 97307726-97307726 on Build GRCh38

View Full Product Details