Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57562065-57562298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG5
Entrez Gene ID
221188
Gene Name
adhesion G protein-coupled receptor G5
Gene Aliases
GPR114, PGR27
UniGene
Hs.187884
Gene Chromosome Location
Chr.16: 57529339-57578182 on Build GRCh38
Transcripts
NM_001304376.2 NM_001318481.1 NM_153837.3 XM_011522949.2 XM_011522950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510498 c.19C>T p.L7F Chr.16: 57562112-57562112 on Build GRCh38
5783869 c.64+10C>A p.? Chr.16: 57562167-57562167 on Build GRCh38
4925588 c.56C>T p.A19V Chr.16: 57562149-57562149 on Build GRCh38
1302059 c.28T>C p.C10R Chr.16: 57562121-57562121 on Build GRCh38
7369912 c.58A>G p.T20A Chr.16: 57562151-57562151 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753840935 Chr.16: 57562207-57562207 on Build GRCh38
rs753066010 Chr.16: 57562188-57562188 on Build GRCh38
rs778514281 Chr.16: 57562139-57562139 on Build GRCh38
rs755662054 Chr.16: 57562099-57562099 on Build GRCh38
rs749941072 Chr.16: 57562098-57562098 on Build GRCh38
rs201729290 Chr.16: 57562105-57562105 on Build GRCh38
rs1004363 Chr.16: 57562087-57562087 on Build GRCh38
rs770952390 Chr.16: 57562165-57562165 on Build GRCh38
rs562904187 Chr.16: 57562245-57562245 on Build GRCh38
rs772253046 Chr.16: 57562148-57562148 on Build GRCh38
rs780891536 Chr.16: 57562090-57562090 on Build GRCh38
rs749199897 Chr.16: 57562122-57562122 on Build GRCh38
rs141637452 Chr.16: 57562146-57562146 on Build GRCh38
rs115266147 Chr.16: 57562151-57562151 on Build GRCh38
rs763345234 Chr.16: 57562189-57562189 on Build GRCh38
rs190137377 Chr.16: 57562227-57562227 on Build GRCh38
rs767122139 Chr.16: 57562193-57562193 on Build GRCh38
rs765714835 Chr.16: 57562184-57562184 on Build GRCh38
rs777123480 Chr.16: 57562166-57562166 on Build GRCh38
rs372612041 Chr.16: 57562208-57562208 on Build GRCh38
rs768578192 Chr.16: 57562133-57562133 on Build GRCh38
rs751234732 Chr.16: 57562086-57562086 on Build GRCh38
rs1056241890 Chr.16: 57562236-57562236 on Build GRCh38
rs755717553 Chr.16: 57562199-57562199 on Build GRCh38
rs892363060 Chr.16: 57562254-57562254 on Build GRCh38
rs750042252 Chr.16: 57562197-57562197 on Build GRCh38
rs763669863 Chr.16: 57562192-57562192 on Build GRCh38
rs184812598 Chr.16: 57562200-57562200 on Build GRCh38
rs557850117 Chr.16: 57562262-57562262 on Build GRCh38
rs554809963 Chr.16: 57562178-57562178 on Build GRCh38
rs1003775674 Chr.16: 57562228-57562228 on Build GRCh38

View Full Product Details