Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144530693-144530966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP39
Entrez Gene ID
80728
Gene Name
Rho GTPase activating protein 39
Gene Aliases
CrGAP, Vilse
UniGene
Hs.148988
Gene Chromosome Location
Chr.8: 144529179-144700520 on Build GRCh38
Transcripts
NM_001308207.1 NM_001308208.1 NM_025251.2 XM_011517308.1 XM_011517309.1 XM_011517312.2 XM_017013870.1 XM_017013871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5998216 c.3140G>A p.R1047H Chr.8: 144530712-144530712 on Build GRCh38
3763204 c.3066C>T p.D1022D Chr.8: 144530786-144530786 on Build GRCh38
6432325 c.3086C>T p.A1029V Chr.8: 144530766-144530766 on Build GRCh38
6603611 c.3115C>G p.R1039G Chr.8: 144530737-144530737 on Build GRCh38
4806048 c.3004C>T p.R1002W Chr.8: 144530848-144530848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555499437 Chr.8: 144530941-144530941 on Build GRCh38
rs769830851 Chr.8: 144530870-144530870 on Build GRCh38
rs759171540 Chr.8: 144530821-144530821 on Build GRCh38
rs754679987 Chr.8: 144530772-144530772 on Build GRCh38
rs762520714 Chr.8: 144530906-144530906 on Build GRCh38
rs751304335 Chr.8: 144530846-144530846 on Build GRCh38
rs773151166 Chr.8: 144530896-144530896 on Build GRCh38
rs763813078 Chr.8: 144530840-144530840 on Build GRCh38
rs143245385 Chr.8: 144530787-144530787 on Build GRCh38
rs1039752869 Chr.8: 144530913-144530913 on Build GRCh38
rs931181843 Chr.8: 144530934-144530934 on Build GRCh38
rs763178671 Chr.8: 144530746-144530746 on Build GRCh38
rs758173664 Chr.8: 144530783-144530783 on Build GRCh38
rs2620648 Chr.8: 144530947-144530947 on Build GRCh38
rs757163896 Chr.8: 144530794-144530794 on Build GRCh38
rs544669862 Chr.8: 144530774-144530774 on Build GRCh38
rs775594759 Chr.8: 144530737-144530737 on Build GRCh38
rs775086545 Chr.8: 144530899-144530899 on Build GRCh38
rs375032023 Chr.8: 144530797-144530797 on Build GRCh38
rs760086265 Chr.8: 144530891-144530891 on Build GRCh38
rs771986069 Chr.8: 144530724-144530724 on Build GRCh38
rs777682905 Chr.8: 144530772-144530773 on Build GRCh38
rs777271175 Chr.8: 144530819-144530819 on Build GRCh38
rs539971387 Chr.8: 144530909-144530909 on Build GRCh38
rs202148580 Chr.8: 144530810-144530810 on Build GRCh38
rs771183983 Chr.8: 144530887-144530887 on Build GRCh38
rs912537790 Chr.8: 144530898-144530898 on Build GRCh38
rs778922705 Chr.8: 144530872-144530872 on Build GRCh38
rs144322342 Chr.8: 144530756-144530756 on Build GRCh38
rs377143451 Chr.8: 144530788-144530788 on Build GRCh38
rs753338775 Chr.8: 144530770-144530770 on Build GRCh38
rs750934223 Chr.8: 144530765-144530765 on Build GRCh38
rs373327760 Chr.8: 144530780-144530780 on Build GRCh38
rs922327170 Chr.8: 144530733-144530733 on Build GRCh38
rs774553358 Chr.8: 144530750-144530750 on Build GRCh38
rs758372999 Chr.8: 144530751-144530752 on Build GRCh38
rs529542732 Chr.8: 144530813-144530813 on Build GRCh38
rs902980404 Chr.8: 144530923-144530923 on Build GRCh38
rs761719118 Chr.8: 144530908-144530908 on Build GRCh38
rs762223990 Chr.8: 144530755-144530755 on Build GRCh38
rs773451836 Chr.8: 144530879-144530879 on Build GRCh38
rs780115976 Chr.8: 144530795-144530795 on Build GRCh38
rs873884 Chr.8: 144530786-144530786 on Build GRCh38
rs765828371 Chr.8: 144530767-144530767 on Build GRCh38
rs781166050 Chr.8: 144530867-144530867 on Build GRCh38
rs772277817 Chr.8: 144530878-144530878 on Build GRCh38
rs755996297 Chr.8: 144530915-144530915 on Build GRCh38
rs777626090 Chr.8: 144530728-144530728 on Build GRCh38
rs746019349 Chr.8: 144530864-144530865 on Build GRCh38
rs550718080 Chr.8: 144530822-144530822 on Build GRCh38
rs566722002 Chr.8: 144530937-144530937 on Build GRCh38
rs757001082 Chr.8: 144530849-144530849 on Build GRCh38
rs551880628 Chr.8: 144530917-144530917 on Build GRCh38
rs756090613 Chr.8: 144530919-144530919 on Build GRCh38
rs752433867 Chr.8: 144530834-144530834 on Build GRCh38
rs1046862485 Chr.8: 144530936-144530936 on Build GRCh38
rs748340606 Chr.8: 144530875-144530875 on Build GRCh38
rs369214772 Chr.8: 144530886-144530886 on Build GRCh38
rs369823775 Chr.8: 144530736-144530736 on Build GRCh38
rs920003683 Chr.8: 144530933-144530933 on Build GRCh38
rs149589076 Chr.8: 144530713-144530713 on Build GRCh38
rs569239939 Chr.8: 144530907-144530907 on Build GRCh38
rs112342135 Chr.8: 144530789-144530789 on Build GRCh38
rs748339987 Chr.8: 144530809-144530809 on Build GRCh38
rs774604745 Chr.8: 144530800-144530800 on Build GRCh38
rs778977574 Chr.8: 144530921-144530921 on Build GRCh38
rs895886873 Chr.8: 144530828-144530828 on Build GRCh38
rs867615474 Chr.8: 144530745-144530745 on Build GRCh38
rs373974961 Chr.8: 144530910-144530910 on Build GRCh38
rs758023189 Chr.8: 144530839-144530839 on Build GRCh38
rs768711526 Chr.8: 144530798-144530798 on Build GRCh38
rs866336176 Chr.8: 144530847-144530847 on Build GRCh38
rs777322640 Chr.8: 144530888-144530888 on Build GRCh38
rs768803401 Chr.8: 144530749-144530749 on Build GRCh38

View Full Product Details