Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113832573-113832800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5448569 c.1044C>A p.P348P Chr.13: 113832672-113832672 on Build GRCh38
7220273 c.943C>T p.R315C Chr.13: 113832773-113832773 on Build GRCh38
432173 c.870G>C p.K290N Chr.13: 113832717-113832717 on Build GRCh38
4409693 c.892G>A p.G298S Chr.13: 113832695-113832695 on Build GRCh38
5448568 c.915C>A p.P305P Chr.13: 113832672-113832672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777752250 Chr.13: 113832679-113832679 on Build GRCh38
rs767132759 Chr.13: 113832708-113832708 on Build GRCh38
rs199643057 Chr.13: 113832736-113832736 on Build GRCh38
rs764065889 Chr.13: 113832756-113832756 on Build GRCh38
rs201605000 Chr.13: 113832760-113832760 on Build GRCh38
rs147632674 Chr.13: 113832691-113832691 on Build GRCh38
rs779039704 Chr.13: 113832604-113832604 on Build GRCh38
rs756029174 Chr.13: 113832674-113832674 on Build GRCh38
rs542438763 Chr.13: 113832672-113832672 on Build GRCh38
rs79109564 Chr.13: 113832725-113832725 on Build GRCh38
rs773435182 Chr.13: 113832780-113832780 on Build GRCh38
rs149144442 Chr.13: 113832692-113832692 on Build GRCh38
rs759006256 Chr.13: 113832667-113832667 on Build GRCh38
rs141710031 Chr.13: 113832763-113832763 on Build GRCh38
rs555163572 Chr.13: 113832665-113832665 on Build GRCh38
rs746086233 Chr.13: 113832701-113832701 on Build GRCh38
rs376986269 Chr.13: 113832727-113832727 on Build GRCh38
rs771176163 Chr.13: 113832603-113832603 on Build GRCh38
rs753633530 Chr.13: 113832681-113832681 on Build GRCh38
rs759198463 Chr.13: 113832738-113832738 on Build GRCh38
rs372518196 Chr.13: 113832772-113832772 on Build GRCh38
rs1038992903 Chr.13: 113832650-113832650 on Build GRCh38
rs577985573 Chr.13: 113832764-113832764 on Build GRCh38
rs200744856 Chr.13: 113832643-113832643 on Build GRCh38
rs375897325 Chr.13: 113832659-113832659 on Build GRCh38
rs147601449 Chr.13: 113832775-113832775 on Build GRCh38
rs1031332459 Chr.13: 113832753-113832753 on Build GRCh38
rs935732068 Chr.13: 113832645-113832645 on Build GRCh38
rs777054303 Chr.13: 113832726-113832726 on Build GRCh38
rs758690045 Chr.13: 113832768-113832768 on Build GRCh38
rs527535980 Chr.13: 113832747-113832747 on Build GRCh38
rs773808304 Chr.13: 113832684-113832684 on Build GRCh38
rs776076149 Chr.13: 113832641-113832641 on Build GRCh38
rs767016321 Chr.13: 113832671-113832671 on Build GRCh38
rs906798049 Chr.13: 113832690-113832690 on Build GRCh38
rs779159581 Chr.13: 113832694-113832694 on Build GRCh38
rs753723766 Chr.13: 113832779-113832779 on Build GRCh38
rs145546824 Chr.13: 113832773-113832773 on Build GRCh38
rs141287368 Chr.13: 113832658-113832658 on Build GRCh38
rs369813911 Chr.13: 113832702-113832702 on Build GRCh38
rs749092015 Chr.13: 113832597-113832597 on Build GRCh38
rs745915511 Chr.13: 113832614-113832614 on Build GRCh38
rs540269924 Chr.13: 113832759-113832759 on Build GRCh38
rs1005804227 Chr.13: 113832635-113832635 on Build GRCh38
rs747155404 Chr.13: 113832714-113832714 on Build GRCh38
rs1018868205 Chr.13: 113832596-113832596 on Build GRCh38
rs1036302663 Chr.13: 113832698-113832698 on Build GRCh38
rs371350136 Chr.13: 113832631-113832631 on Build GRCh38
rs371599789 Chr.13: 113832608-113832608 on Build GRCh38
rs373401212 Chr.13: 113832776-113832776 on Build GRCh38
rs200207558 Chr.13: 113832737-113832737 on Build GRCh38
rs376441448 Chr.13: 113832741-113832741 on Build GRCh38
rs769346633 Chr.13: 113832655-113832655 on Build GRCh38
rs779402707 Chr.13: 113832774-113832774 on Build GRCh38
rs374837053 Chr.13: 113832682-113832682 on Build GRCh38
rs200560351 Chr.13: 113832771-113832771 on Build GRCh38
rs760214185 Chr.13: 113832755-113832755 on Build GRCh38
rs145302934 Chr.13: 113832664-113832664 on Build GRCh38
rs772313363 Chr.13: 113832619-113832619 on Build GRCh38

View Full Product Details