Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22172067-22172298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035296 c.895_930del36 p.L300_S311del12 Chr.18: 22172039-22172074 on Build GRCh38
6036397 c.968_985del18 p.H328_H333delHHHHHH Chr.18: 22172112-22172129 on Build GRCh38
5513999 c.968_969insACC p.Y323>* Chr.18: 22172112-22172113 on Build GRCh38
6383213 c.1069C>A p.P357T Chr.18: 22172213-22172213 on Build GRCh38
4888561 c.1069C>T p.P357S Chr.18: 22172213-22172213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922570984 Chr.18: 22172252-22172252 on Build GRCh38
rs142601402 Chr.18: 22172171-22172171 on Build GRCh38
rs797045593 Chr.18: 22172231-22172231 on Build GRCh38
rs587780342 Chr.18: 22172112-22172114 on Build GRCh38
rs768797991 Chr.18: 22172276-22172276 on Build GRCh38
rs763204851 Chr.18: 22172141-22172143 on Build GRCh38
rs587777711 Chr.18: 22172216-22172216 on Build GRCh38
rs878854756 Chr.18: 22172137-22172142 on Build GRCh38
rs1018392814 Chr.18: 22172188-22172188 on Build GRCh38
rs964471147 Chr.18: 22172192-22172192 on Build GRCh38
rs963001157 Chr.18: 22172112-22172120 on Build GRCh38
rs1011495923 Chr.18: 22172133-22172133 on Build GRCh38
rs976781879 Chr.18: 22172194-22172194 on Build GRCh38
rs954640705 Chr.18: 22172267-22172267 on Build GRCh38
rs970282660 Chr.18: 22172086-22172086 on Build GRCh38
rs746206001 Chr.18: 22172128-22172128 on Build GRCh38
rs749692988 Chr.18: 22172265-22172265 on Build GRCh38
rs973074328 Chr.18: 22172131-22172131 on Build GRCh38
rs562588574 Chr.18: 22172111-22172112 on Build GRCh38
rs780646014 Chr.18: 22172219-22172219 on Build GRCh38
rs1017227591 Chr.18: 22172112-22172117 on Build GRCh38
rs770099946 Chr.18: 22172154-22172154 on Build GRCh38
rs1002187848 Chr.18: 22172089-22172089 on Build GRCh38
rs797045180 Chr.18: 22172215-22172215 on Build GRCh38

View Full Product Details