Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177280870-177281105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP2
Entrez Gene ID
57795
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
DBCCR1L2, FAM5B
UniGene
Hs.495918
Gene Chromosome Location
Chr.1: 177170922-177282422 on Build GRCh38
Transcripts
NM_021165.3 XM_005245379.1 XM_011509826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260811 c.1797G>A p.E599E Chr.1: 177280973-177280973 on Build GRCh38
6123081 c.1749G>T p.E583D Chr.1: 177280925-177280925 on Build GRCh38
4828554 c.1793C>G p.S598C Chr.1: 177280969-177280969 on Build GRCh38
6534057 c.1779T>G p.F593L Chr.1: 177280955-177280955 on Build GRCh38
219450 c.1775C>A p.P592H Chr.1: 177280951-177280951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753028699 Chr.1: 177281039-177281039 on Build GRCh38
rs771967341 Chr.1: 177280997-177280997 on Build GRCh38
rs765386717 Chr.1: 177281032-177281032 on Build GRCh38
rs148283069 Chr.1: 177280959-177280960 on Build GRCh38
rs758546554 Chr.1: 177281046-177281046 on Build GRCh38
rs142394323 Chr.1: 177280960-177280961 on Build GRCh38
rs759731438 Chr.1: 177281028-177281028 on Build GRCh38
rs755061077 Chr.1: 177280928-177280928 on Build GRCh38
rs971088191 Chr.1: 177280917-177280917 on Build GRCh38
rs775161210 Chr.1: 177280937-177280960 on Build GRCh38
rs751886595 Chr.1: 177281075-177281075 on Build GRCh38
rs183912741 Chr.1: 177281061-177281061 on Build GRCh38
rs147592131 Chr.1: 177280979-177280979 on Build GRCh38
rs766249875 Chr.1: 177281011-177281011 on Build GRCh38
rs559412885 Chr.1: 177280944-177280944 on Build GRCh38
rs145369948 Chr.1: 177280904-177280904 on Build GRCh38
rs768120673 Chr.1: 177280960-177280960 on Build GRCh38
rs868647928 Chr.1: 177280956-177280956 on Build GRCh38
rs748351328 Chr.1: 177280933-177280933 on Build GRCh38
rs760459965 Chr.1: 177281010-177281010 on Build GRCh38
rs200578567 Chr.1: 177281079-177281079 on Build GRCh38
rs753787662 Chr.1: 177281027-177281027 on Build GRCh38
rs772887548 Chr.1: 177280998-177280998 on Build GRCh38
rs747622333 Chr.1: 177280948-177280948 on Build GRCh38
rs761440262 Chr.1: 177280995-177280995 on Build GRCh38
rs993065363 Chr.1: 177281044-177281044 on Build GRCh38
rs758886283 Chr.1: 177280943-177280943 on Build GRCh38
rs946224963 Chr.1: 177281065-177281065 on Build GRCh38
rs779310557 Chr.1: 177280930-177280930 on Build GRCh38
rs191738799 Chr.1: 177280957-177280957 on Build GRCh38
rs201494475 Chr.1: 177280978-177280978 on Build GRCh38
rs796395753 Chr.1: 177281026-177281026 on Build GRCh38
rs777143325 Chr.1: 177280955-177280955 on Build GRCh38
rs771351231 Chr.1: 177280953-177280953 on Build GRCh38
rs774037708 Chr.1: 177280967-177280967 on Build GRCh38

View Full Product Details