Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32441795-32442060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM7
Entrez Gene ID
112616
Gene Name
CKLF like MARVEL transmembrane domain containing 7
Gene Aliases
CKLFSF7
UniGene
Hs.440494
Gene Chromosome Location
Chr.3: 32391671-32455528 on Build GRCh38
Transcripts
NM_138410.3 NM_181472.2 XM_011533319.2 XM_017005646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188317 c.326C>T p.P109L Chr.3: 32442006-32442006 on Build GRCh38
6237795 c.282C>A p.V94V Chr.3: 32441962-32441962 on Build GRCh38
6258960 c.224G>T p.S75I Chr.3: 32441904-32441904 on Build GRCh38
4430995 c.267C>T p.L89L Chr.3: 32441947-32441947 on Build GRCh38
6097485 c.164C>A p.T55N Chr.3: 32441844-32441844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780296829 Chr.3: 32441930-32441930 on Build GRCh38
rs372474802 Chr.3: 32442025-32442025 on Build GRCh38
rs1057302127 Chr.3: 32441947-32441947 on Build GRCh38
rs769547195 Chr.3: 32441851-32441851 on Build GRCh38
rs1007434213 Chr.3: 32441883-32441883 on Build GRCh38
rs751108343 Chr.3: 32441865-32441865 on Build GRCh38
rs149342668 Chr.3: 32441833-32441833 on Build GRCh38
rs866824839 Chr.3: 32442006-32442006 on Build GRCh38
rs779250384 Chr.3: 32441900-32441900 on Build GRCh38
rs752362450 Chr.3: 32441870-32441870 on Build GRCh38
rs759152912 Chr.3: 32441967-32441967 on Build GRCh38
rs748994901 Chr.3: 32441965-32441965 on Build GRCh38
rs754525393 Chr.3: 32442013-32442013 on Build GRCh38
rs532611688 Chr.3: 32441896-32441896 on Build GRCh38
rs761645113 Chr.3: 32441848-32441848 on Build GRCh38
rs745984521 Chr.3: 32441911-32441911 on Build GRCh38
rs778191561 Chr.3: 32441872-32441872 on Build GRCh38
rs144740563 Chr.3: 32441929-32441929 on Build GRCh38
rs200217502 Chr.3: 32441857-32441857 on Build GRCh38
rs760421593 Chr.3: 32441975-32441975 on Build GRCh38
rs774130476 Chr.3: 32441966-32441966 on Build GRCh38
rs767127146 Chr.3: 32441972-32441972 on Build GRCh38
rs142565885 Chr.3: 32441983-32441983 on Build GRCh38
rs190577874 Chr.3: 32442041-32442041 on Build GRCh38
rs948961021 Chr.3: 32441822-32441822 on Build GRCh38
rs17029526 Chr.3: 32441866-32441866 on Build GRCh38
rs377268191 Chr.3: 32441948-32441948 on Build GRCh38
rs762960087 Chr.3: 32441862-32441862 on Build GRCh38
rs201511433 Chr.3: 32441878-32441878 on Build GRCh38
rs117098616 Chr.3: 32441982-32441982 on Build GRCh38
rs758541188 Chr.3: 32442005-32442005 on Build GRCh38
rs756618154 Chr.3: 32441871-32441871 on Build GRCh38
rs765824035 Chr.3: 32441984-32441984 on Build GRCh38
rs370800472 Chr.3: 32442010-32442010 on Build GRCh38
rs994617278 Chr.3: 32442026-32442026 on Build GRCh38
rs773063152 Chr.3: 32441957-32441957 on Build GRCh38
rs766159956 Chr.3: 32441864-32441864 on Build GRCh38
rs1017441804 Chr.3: 32441897-32441897 on Build GRCh38
rs775643254 Chr.3: 32441846-32441846 on Build GRCh38
rs373532397 Chr.3: 32441869-32441869 on Build GRCh38
rs980324617 Chr.3: 32441830-32441830 on Build GRCh38
rs745871953 Chr.3: 32441814-32441814 on Build GRCh38
rs775344390 Chr.3: 32441974-32441974 on Build GRCh38
rs763589738 Chr.3: 32441981-32441981 on Build GRCh38
rs747217676 Chr.3: 32441938-32441938 on Build GRCh38

View Full Product Details