Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237094007-237094280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS8
Entrez Gene ID
10920
Gene Name
COP9 signalosome subunit 8
Gene Aliases
COP9, CSN8, SGN8
UniGene
Hs.531713
Gene Chromosome Location
Chr.2: 237085441-237098846 on Build GRCh38
Transcripts
NM_006710.4 NM_198189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4981818 c.353T>A p.F118Y Chr.2: 237094111-237094111 on Build GRCh38
4956896 c.347G>T p.R116L Chr.2: 237094105-237094105 on Build GRCh38
3051214 c.391G>A p.D131N Chr.2: 237094149-237094149 on Build GRCh38
6281770 c.343A>G p.R115G Chr.2: 237094101-237094101 on Build GRCh38
3051213 c.349G>A p.A117T Chr.2: 237094107-237094107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79436713 Chr.2: 237094236-237094236 on Build GRCh38
rs777181775 Chr.2: 237094090-237094090 on Build GRCh38
rs759959050 Chr.2: 237094187-237094187 on Build GRCh38
rs73999619 Chr.2: 237094206-237094206 on Build GRCh38
rs779821641 Chr.2: 237094247-237094247 on Build GRCh38
rs780058598 Chr.2: 237094149-237094149 on Build GRCh38
rs776105885 Chr.2: 237094195-237094195 on Build GRCh38
rs774842550 Chr.2: 237094060-237094061 on Build GRCh38
rs545822553 Chr.2: 237094222-237094222 on Build GRCh38
rs1031626586 Chr.2: 237094093-237094093 on Build GRCh38
rs151029276 Chr.2: 237094125-237094125 on Build GRCh38
rs778513456 Chr.2: 237094162-237094162 on Build GRCh38
rs747854981 Chr.2: 237094163-237094163 on Build GRCh38
rs750418832 Chr.2: 237094048-237094059 on Build GRCh38
rs765778911 Chr.2: 237094106-237094106 on Build GRCh38
rs771279491 Chr.2: 237094171-237094171 on Build GRCh38
rs188786181 Chr.2: 237094194-237094194 on Build GRCh38
rs768191872 Chr.2: 237094151-237094151 on Build GRCh38
rs779368853 Chr.2: 237094039-237094039 on Build GRCh38
rs147957647 Chr.2: 237094136-237094136 on Build GRCh38
rs938433098 Chr.2: 237094196-237094196 on Build GRCh38
rs748848460 Chr.2: 237094050-237094050 on Build GRCh38
rs533865440 Chr.2: 237094148-237094148 on Build GRCh38
rs149840872 Chr.2: 237094130-237094130 on Build GRCh38
rs750140815 Chr.2: 237094230-237094230 on Build GRCh38
rs965405157 Chr.2: 237094080-237094080 on Build GRCh38
rs374366971 Chr.2: 237094239-237094239 on Build GRCh38
rs202129594 Chr.2: 237094129-237094129 on Build GRCh38
rs367631675 Chr.2: 237094145-237094145 on Build GRCh38
rs768244964 Chr.2: 237094051-237094051 on Build GRCh38
rs760027685 Chr.2: 237094105-237094105 on Build GRCh38
rs373917449 Chr.2: 237094224-237094224 on Build GRCh38
rs755964282 Chr.2: 237094246-237094246 on Build GRCh38
rs764329716 Chr.2: 237094207-237094207 on Build GRCh38
rs373998005 Chr.2: 237094208-237094208 on Build GRCh38
rs751881755 Chr.2: 237094134-237094134 on Build GRCh38
rs760994846 Chr.2: 237094076-237094076 on Build GRCh38
rs767797885 Chr.2: 237094223-237094223 on Build GRCh38
rs780919409 Chr.2: 237094140-237094140 on Build GRCh38
rs752671265 Chr.2: 237094107-237094107 on Build GRCh38
rs373807335 Chr.2: 237094084-237094084 on Build GRCh38
rs760292225 Chr.2: 237094165-237094173 on Build GRCh38
rs753819120 Chr.2: 237094248-237094248 on Build GRCh38
rs751829342 Chr.2: 237094220-237094220 on Build GRCh38
rs776793969 Chr.2: 237094184-237094184 on Build GRCh38
rs115416168 Chr.2: 237094067-237094067 on Build GRCh38

View Full Product Details