Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49860272-49860513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKV
Entrez Gene ID
79012
Gene Name
CaM kinase like vesicle associated
Gene Aliases
1G5, VACAMKL
UniGene
Hs.145156
Gene Chromosome Location
Chr.3: 49857981-49870222 on Build GRCh38
Transcripts
NM_001320147.1 NM_024046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
360922 c.820C>A p.R274R Chr.3: 49860505-49860505 on Build GRCh38
5399620 c.814G>A p.D272N Chr.3: 49860511-49860511 on Build GRCh38
6371137 c.839C>T p.A280V Chr.3: 49860486-49860486 on Build GRCh38
3595425 c.825C>T p.I275I Chr.3: 49860500-49860500 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs972743709 Chr.3: 49860439-49860439 on Build GRCh38
rs745483940 Chr.3: 49860464-49860464 on Build GRCh38
rs770578942 Chr.3: 49860453-49860453 on Build GRCh38
rs751741882 Chr.3: 49860299-49860299 on Build GRCh38
rs944287748 Chr.3: 49860473-49860473 on Build GRCh38
rs2681780 Chr.3: 49860397-49860397 on Build GRCh38
rs781225331 Chr.3: 49860421-49860421 on Build GRCh38
rs1005709394 Chr.3: 49860344-49860344 on Build GRCh38
rs372936283 Chr.3: 49860450-49860450 on Build GRCh38
rs774315831 Chr.3: 49860456-49860456 on Build GRCh38
rs911505494 Chr.3: 49860428-49860428 on Build GRCh38
rs756072830 Chr.3: 49860445-49860445 on Build GRCh38
rs889989595 Chr.3: 49860329-49860329 on Build GRCh38
rs56071455 Chr.3: 49860488-49860488 on Build GRCh38
rs952983377 Chr.3: 49860377-49860377 on Build GRCh38
rs11714822 Chr.3: 49860305-49860305 on Build GRCh38
rs748232430 Chr.3: 49860437-49860437 on Build GRCh38
rs537943777 Chr.3: 49860300-49860300 on Build GRCh38
rs918737031 Chr.3: 49860485-49860485 on Build GRCh38
rs770211194 Chr.3: 49860345-49860345 on Build GRCh38

View Full Product Details