Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33132488-33132761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAP
Entrez Gene ID
10491
Gene Name
cartilage associated protein
Gene Aliases
CASP, LEPREL3, OI7, P3H5
UniGene
Hs.517888
Gene Chromosome Location
Chr.3: 33113958-33147773 on Build GRCh38
Transcripts
NM_006371.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002658 c.1044G>C p.S348S Chr.3: 33132676-33132676 on Build GRCh38
6097477 c.928G>C p.D310H Chr.3: 33132560-33132560 on Build GRCh38
6635736 c.932T>A p.L311Q Chr.3: 33132564-33132564 on Build GRCh38
4674789 c.962A>G p.Y321C Chr.3: 33132594-33132594 on Build GRCh38
149349 c.1032T>G p.T344T Chr.3: 33132664-33132664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115198029 Chr.3: 33132671-33132671 on Build GRCh38
rs201566559 Chr.3: 33132727-33132727 on Build GRCh38
rs137853947 Chr.3: 33132553-33132553 on Build GRCh38
rs545368905 Chr.3: 33132675-33132675 on Build GRCh38
rs116821774 Chr.3: 33132608-33132608 on Build GRCh38
rs768954904 Chr.3: 33132652-33132652 on Build GRCh38
rs1135128 Chr.3: 33132676-33132676 on Build GRCh38
rs908834247 Chr.3: 33132678-33132678 on Build GRCh38
rs1011445335 Chr.3: 33132577-33132577 on Build GRCh38
rs201997191 Chr.3: 33132580-33132580 on Build GRCh38
rs756963885 Chr.3: 33132614-33132614 on Build GRCh38
rs757967789 Chr.3: 33132618-33132618 on Build GRCh38
rs771061593 Chr.3: 33132670-33132670 on Build GRCh38
rs780660572 Chr.3: 33132524-33132524 on Build GRCh38
rs753670810 Chr.3: 33132683-33132683 on Build GRCh38
rs1135127 Chr.3: 33132664-33132664 on Build GRCh38
rs757797834 Chr.3: 33132730-33132730 on Build GRCh38
rs375233411 Chr.3: 33132520-33132520 on Build GRCh38
rs769801562 Chr.3: 33132579-33132579 on Build GRCh38
rs772446555 Chr.3: 33132523-33132523 on Build GRCh38
rs573914818 Chr.3: 33132594-33132594 on Build GRCh38
rs962668670 Chr.3: 33132555-33132555 on Build GRCh38
rs114178925 Chr.3: 33132585-33132585 on Build GRCh38
rs773519852 Chr.3: 33132667-33132667 on Build GRCh38
rs765931039 Chr.3: 33132736-33132736 on Build GRCh38
rs778682499 Chr.3: 33132615-33132615 on Build GRCh38
rs745448109 Chr.3: 33132521-33132521 on Build GRCh38
rs747344179 Chr.3: 33132548-33132548 on Build GRCh38
rs1025718805 Chr.3: 33132564-33132564 on Build GRCh38
rs752708438 Chr.3: 33132604-33132604 on Build GRCh38
rs773163098 Chr.3: 33132586-33132586 on Build GRCh38
rs778289479 Chr.3: 33132518-33132518 on Build GRCh38
rs111525897 Chr.3: 33132560-33132560 on Build GRCh38
rs974643671 Chr.3: 33132688-33132688 on Build GRCh38
rs773524867 Chr.3: 33132637-33132637 on Build GRCh38
rs762897204 Chr.3: 33132669-33132669 on Build GRCh38
rs749879935 Chr.3: 33132617-33132617 on Build GRCh38
rs763492697 Chr.3: 33132559-33132559 on Build GRCh38
rs753473622 Chr.3: 33132610-33132610 on Build GRCh38
rs760493228 Chr.3: 33132607-33132607 on Build GRCh38
rs369417539 Chr.3: 33132716-33132716 on Build GRCh38
rs762039541 Chr.3: 33132562-33132562 on Build GRCh38
rs747540938 Chr.3: 33132650-33132650 on Build GRCh38
rs781545232 Chr.3: 33132654-33132654 on Build GRCh38
rs761750861 Chr.3: 33132711-33132711 on Build GRCh38
rs757988143 Chr.3: 33132551-33132551 on Build GRCh38
rs969841258 Chr.3: 33132640-33132640 on Build GRCh38
rs780385904 Chr.3: 33132638-33132638 on Build GRCh38

View Full Product Details