Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45040128-45040401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASRP
Entrez Gene ID
11129
Gene Name
CLK4 associating serine/arginine rich protein
Gene Aliases
CLASP, SFRS16, SWAP2
UniGene
Hs.466917
Gene Chromosome Location
Chr.19: 45039040-45070956 on Build GRCh38
Transcripts
NM_001278439.1 NM_007056.2 NR_103529.1 XM_011526396.2 XM_011526397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397022 c.52G>A p.D18N Chr.19: 45040264-45040264 on Build GRCh38
6276263 c.41G>T p.G14V Chr.19: 45040253-45040253 on Build GRCh38
6629751 c.64A>G p.R22G Chr.19: 45040276-45040276 on Build GRCh38
4920775 c.51C>T p.V17V Chr.19: 45040263-45040263 on Build GRCh38
7313014 c.69G>T p.A23A Chr.19: 45040281-45040281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976155536 Chr.19: 45040336-45040336 on Build GRCh38
rs984805952 Chr.19: 45040359-45040359 on Build GRCh38
rs757987658 Chr.19: 45040278-45040278 on Build GRCh38
rs763107800 Chr.19: 45040181-45040181 on Build GRCh38
rs376408519 Chr.19: 45040220-45040220 on Build GRCh38
rs764009712 Chr.19: 45040205-45040205 on Build GRCh38
rs757003701 Chr.19: 45040263-45040263 on Build GRCh38
rs919350960 Chr.19: 45040340-45040340 on Build GRCh38
rs911037546 Chr.19: 45040189-45040189 on Build GRCh38
rs986355993 Chr.19: 45040281-45040281 on Build GRCh38
rs781754330 Chr.19: 45040334-45040334 on Build GRCh38
rs746486921 Chr.19: 45040302-45040302 on Build GRCh38
rs751599790 Chr.19: 45040184-45040184 on Build GRCh38
rs750030224 Chr.19: 45040275-45040275 on Build GRCh38
rs752716804 Chr.19: 45040348-45040352 on Build GRCh38
rs770236460 Chr.19: 45040343-45040343 on Build GRCh38
rs768329475 Chr.19: 45040324-45040324 on Build GRCh38
rs370663852 Chr.19: 45040201-45040201 on Build GRCh38
rs773528960 Chr.19: 45040164-45040164 on Build GRCh38
rs935261389 Chr.19: 45040335-45040335 on Build GRCh38
rs748698762 Chr.19: 45040339-45040339 on Build GRCh38
rs779823314 Chr.19: 45040296-45040296 on Build GRCh38
rs572924030 Chr.19: 45040183-45040183 on Build GRCh38
rs753825106 Chr.19: 45040213-45040213 on Build GRCh38
rs376349112 Chr.19: 45040355-45040355 on Build GRCh38

View Full Product Details