Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22892827-22893060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB2
Entrez Gene ID
2048
Gene Name
EPH receptor B2
Gene Aliases
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5
UniGene
Hs.380705
Gene Chromosome Location
Chr.1: 22710770-22921500 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4877622 c.1532G>A p.R511H Chr.1: 22892987-22892987 on Build GRCh38
5911314 c.1429G>A p.E477K Chr.1: 22892884-22892884 on Build GRCh38
131753 c.1535C>T p.T512I Chr.1: 22892990-22892990 on Build GRCh38
4877624 c.1517G>A p.R506H Chr.1: 22892987-22892987 on Build GRCh38
3721651 c.1476C>A p.G492G Chr.1: 22892946-22892946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780502252 Chr.1: 22892875-22892875 on Build GRCh38
rs367743248 Chr.1: 22892997-22892997 on Build GRCh38
rs756162736 Chr.1: 22892909-22892909 on Build GRCh38
rs2746331 Chr.1: 22892851-22892851 on Build GRCh38
rs543651225 Chr.1: 22892937-22892937 on Build GRCh38
rs370664820 Chr.1: 22892931-22892931 on Build GRCh38
rs762242282 Chr.1: 22892966-22892966 on Build GRCh38
rs34273063 Chr.1: 22893003-22893003 on Build GRCh38
rs147223001 Chr.1: 22892958-22892958 on Build GRCh38
rs309472 Chr.1: 22893000-22893000 on Build GRCh38
rs759303586 Chr.1: 22892986-22892986 on Build GRCh38
rs751125040 Chr.1: 22892852-22892852 on Build GRCh38
rs754547916 Chr.1: 22892862-22892862 on Build GRCh38
rs777155810 Chr.1: 22892959-22892959 on Build GRCh38
rs953288463 Chr.1: 22893016-22893016 on Build GRCh38
rs973792941 Chr.1: 22893024-22893024 on Build GRCh38
rs748376698 Chr.1: 22893015-22893015 on Build GRCh38
rs369387828 Chr.1: 22893007-22893007 on Build GRCh38
rs765600046 Chr.1: 22892976-22892976 on Build GRCh38
rs761300345 Chr.1: 22892898-22892898 on Build GRCh38
rs762189253 Chr.1: 22892901-22892901 on Build GRCh38
rs777829293 Chr.1: 22892928-22892928 on Build GRCh38
rs200935672 Chr.1: 22892878-22892878 on Build GRCh38
rs774732880 Chr.1: 22892887-22892887 on Build GRCh38
rs772198021 Chr.1: 22892893-22892893 on Build GRCh38
rs764578709 Chr.1: 22892900-22892900 on Build GRCh38
rs749185690 Chr.1: 22892930-22892930 on Build GRCh38
rs745644967 Chr.1: 22893012-22893012 on Build GRCh38
rs772170964 Chr.1: 22892945-22892945 on Build GRCh38
rs867115042 Chr.1: 22892920-22892920 on Build GRCh38
rs780164151 Chr.1: 22893019-22893019 on Build GRCh38
rs759633255 Chr.1: 22892891-22892891 on Build GRCh38
rs200298496 Chr.1: 22892886-22892886 on Build GRCh38
rs201691912 Chr.1: 22892938-22892938 on Build GRCh38
rs775705074 Chr.1: 22892955-22892955 on Build GRCh38
rs200675482 Chr.1: 22892879-22892879 on Build GRCh38
rs138296617 Chr.1: 22892905-22892906 on Build GRCh38
rs142703174 Chr.1: 22892991-22892991 on Build GRCh38
rs531840904 Chr.1: 22892992-22892992 on Build GRCh38
rs116848191 Chr.1: 22892905-22892905 on Build GRCh38
rs576616864 Chr.1: 22892902-22892902 on Build GRCh38
rs775374349 Chr.1: 22892896-22892896 on Build GRCh38
rs867477730 Chr.1: 22892990-22892990 on Build GRCh38
rs773439627 Chr.1: 22892981-22892981 on Build GRCh38
rs373865135 Chr.1: 22892956-22892956 on Build GRCh38
rs138075206 Chr.1: 22892884-22892884 on Build GRCh38
rs767048571 Chr.1: 22892987-22892987 on Build GRCh38
rs752147967 Chr.1: 22892907-22892907 on Build GRCh38

View Full Product Details