Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105468252-105468481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
UniGene
Hs.136901
Gene Chromosome Location
Chr.9: 105442183-105552433 on Build GRCh38
Transcripts
NM_001145313.2 NM_001287191.1 NM_001287192.1 NM_031919.4 XM_005252254.2 XM_005252257.3 XM_005252260.3 XM_011519077.2 XM_011519078.2 XM_011519079.2 XM_017015182.1 XM_017015183.1 XM_017015184.1 XM_017015185.1 XM_017015186.1 XM_017015187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368881 c.287G>T p.S96I Chr.9: 105468272-105468272 on Build GRCh38
1103563 c.306G>T p.K102N Chr.9: 105468291-105468291 on Build GRCh38
6582477 c.281A>G p.K94R Chr.9: 105468266-105468266 on Build GRCh38
1103564 c.210G>T p.K70N Chr.9: 105468291-105468291 on Build GRCh38
3788133 c.279A>G p.V93V Chr.9: 105468264-105468264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200324817 Chr.9: 105468304-105468304 on Build GRCh38
rs78265177 Chr.9: 105468385-105468385 on Build GRCh38
rs10991664 Chr.9: 105468428-105468428 on Build GRCh38
rs898868338 Chr.9: 105468411-105468411 on Build GRCh38
rs773699591 Chr.9: 105468293-105468293 on Build GRCh38
rs1007836221 Chr.9: 105468397-105468397 on Build GRCh38
rs139888606 Chr.9: 105468392-105468392 on Build GRCh38
rs958054186 Chr.9: 105468434-105468434 on Build GRCh38
rs537092448 Chr.9: 105468334-105468334 on Build GRCh38
rs145969512 Chr.9: 105468366-105468366 on Build GRCh38
rs757479387 Chr.9: 105468353-105468353 on Build GRCh38
rs374590061 Chr.9: 105468344-105468344 on Build GRCh38
rs1018253292 Chr.9: 105468400-105468400 on Build GRCh38
rs373156077 Chr.9: 105468315-105468315 on Build GRCh38
rs950475924 Chr.9: 105468438-105468438 on Build GRCh38
rs572984813 Chr.9: 105468402-105468402 on Build GRCh38
rs1013738216 Chr.9: 105468360-105468360 on Build GRCh38
rs746090809 Chr.9: 105468431-105468431 on Build GRCh38
rs538612432 Chr.9: 105468305-105468305 on Build GRCh38

View Full Product Details