Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70248864-70249101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN3
Entrez Gene ID
1448
Gene Name
casein kappa
Gene Aliases
CNS10, CSN10, CSNK, KCA
UniGene
Hs.54415
Gene Chromosome Location
Chr.4: 70238370-70251432 on Build GRCh38
Transcripts
NM_005212.2 XM_011531620.2 XM_017007761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125730 c.154C>T p.P52S Chr.4: 70249064-70249064 on Build GRCh38
70321 c.154C>G p.P52A Chr.4: 70249064-70249064 on Build GRCh38
3605416 c.120G>A p.Q40Q Chr.4: 70249030-70249030 on Build GRCh38
7151308 c.175G>C p.G59R Chr.4: 70249085-70249085 on Build GRCh38
7203256 c.88-6T>C p.? Chr.4: 70248992-70248992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762332807 Chr.4: 70248956-70248956 on Build GRCh38
rs763832452 Chr.4: 70248971-70248971 on Build GRCh38
rs754134518 Chr.4: 70249055-70249055 on Build GRCh38
rs765608351 Chr.4: 70248950-70248950 on Build GRCh38
rs901000680 Chr.4: 70248906-70248906 on Build GRCh38
rs769300259 Chr.4: 70248986-70248990 on Build GRCh38
rs555054114 Chr.4: 70249051-70249051 on Build GRCh38
rs746172010 Chr.4: 70249063-70249063 on Build GRCh38
rs781703837 Chr.4: 70249058-70249058 on Build GRCh38
rs267600232 Chr.4: 70249050-70249050 on Build GRCh38
rs535342081 Chr.4: 70248985-70248986 on Build GRCh38
rs958720320 Chr.4: 70248994-70248994 on Build GRCh38
rs778186341 Chr.4: 70249030-70249030 on Build GRCh38
rs761732867 Chr.4: 70248986-70248986 on Build GRCh38
rs758357993 Chr.4: 70249026-70249026 on Build GRCh38
rs57584424 Chr.4: 70248969-70248971 on Build GRCh38
rs775814128 Chr.4: 70249066-70249066 on Build GRCh38
rs201750233 Chr.4: 70248986-70248986 on Build GRCh38
rs1025915693 Chr.4: 70248914-70248914 on Build GRCh38
rs767296016 Chr.4: 70248985-70248985 on Build GRCh38
rs916745829 Chr.4: 70249043-70249043 on Build GRCh38
rs746819893 Chr.4: 70248985-70248985 on Build GRCh38
rs3836687 Chr.4: 70248960-70248962 on Build GRCh38
rs57550378 Chr.4: 70248986-70248987 on Build GRCh38
rs1024534620 Chr.4: 70249037-70249037 on Build GRCh38
rs770368585 Chr.4: 70249064-70249064 on Build GRCh38
rs983549260 Chr.4: 70248977-70248977 on Build GRCh38
rs997998186 Chr.4: 70248913-70248913 on Build GRCh38
rs771485308 Chr.4: 70249054-70249054 on Build GRCh38
rs1027660118 Chr.4: 70248921-70248922 on Build GRCh38
rs1039619303 Chr.4: 70248888-70248888 on Build GRCh38
rs747200394 Chr.4: 70249045-70249045 on Build GRCh38
rs112171751 Chr.4: 70248980-70248980 on Build GRCh38
rs779787162 Chr.4: 70249012-70249012 on Build GRCh38
rs991729529 Chr.4: 70249024-70249024 on Build GRCh38
rs751193336 Chr.4: 70248979-70248979 on Build GRCh38
rs756090036 Chr.4: 70248988-70248988 on Build GRCh38
rs367828250 Chr.4: 70249022-70249022 on Build GRCh38
rs775073171 Chr.4: 70248952-70248952 on Build GRCh38
rs397831640 Chr.4: 70248993-70248994 on Build GRCh38

View Full Product Details