Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43625768-43626023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM4
Entrez Gene ID
199731
Gene Name
cell adhesion molecule 4
Gene Aliases
IGSF4C, NECL4, Necl-4, TSLL2, synCAM4
UniGene
Hs.370984
Gene Chromosome Location
Chr.19: 43622368-43641984 on Build GRCh38
Transcripts
NM_145296.1 XM_005258620.2 XM_017026452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866461 c.694C>A p.Q232K Chr.19: 43625972-43625972 on Build GRCh38
5464607 c.755+5G>A p.? Chr.19: 43625906-43625906 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774547485 Chr.19: 43625994-43625994 on Build GRCh38
rs985610485 Chr.19: 43625836-43625836 on Build GRCh38
rs184317257 Chr.19: 43625818-43625818 on Build GRCh38
rs776596557 Chr.19: 43625868-43625868 on Build GRCh38
rs756541498 Chr.19: 43625970-43625970 on Build GRCh38
rs781460070 Chr.19: 43625961-43625961 on Build GRCh38
rs748570389 Chr.19: 43625969-43625969 on Build GRCh38
rs955090425 Chr.19: 43625819-43625819 on Build GRCh38
rs910981891 Chr.19: 43625848-43625848 on Build GRCh38
rs763858251 Chr.19: 43625866-43625866 on Build GRCh38
rs920819568 Chr.19: 43625922-43625922 on Build GRCh38
rs1037893431 Chr.19: 43625921-43625921 on Build GRCh38
rs771052119 Chr.19: 43625874-43625884 on Build GRCh38
rs749911069 Chr.19: 43625880-43625880 on Build GRCh38
rs766071297 Chr.19: 43625946-43625946 on Build GRCh38
rs760445312 Chr.19: 43625863-43625863 on Build GRCh38
rs567666364 Chr.19: 43625978-43625978 on Build GRCh38
rs551608169 Chr.19: 43625838-43625838 on Build GRCh38
rs571693720 Chr.19: 43625851-43625851 on Build GRCh38
rs373219315 Chr.19: 43625888-43625888 on Build GRCh38
rs895804211 Chr.19: 43625987-43625987 on Build GRCh38
rs749312086 Chr.19: 43625991-43625991 on Build GRCh38
rs751323866 Chr.19: 43625947-43625947 on Build GRCh38
rs34246023 Chr.19: 43625993-43625993 on Build GRCh38
rs1047097385 Chr.19: 43625812-43625812 on Build GRCh38
rs376182125 Chr.19: 43625952-43625952 on Build GRCh38
rs765240877 Chr.19: 43625878-43625878 on Build GRCh38
rs1014289603 Chr.19: 43625989-43625989 on Build GRCh38
rs527716544 Chr.19: 43625870-43625870 on Build GRCh38

View Full Product Details