Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203246594-203246829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP20A1
Entrez Gene ID
57404
Gene Name
cytochrome P450 family 20 subfamily A member 1
Gene Aliases
CYP-M
UniGene
Hs.446065
Gene Chromosome Location
Chr.2: 203238441-203305840 on Build GRCh38
Transcripts
NM_177538.2 XM_005246698.3 XM_005246699.3 XM_005246701.3 XM_006712644.3 XM_006712645.3 XM_017004555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3576499 c.136C>T p.P46S Chr.2: 203246768-203246768 on Build GRCh38
1015115 c.130A>C p.N44H Chr.2: 203246762-203246762 on Build GRCh38
4465133 c.136C>G p.P46A Chr.2: 203246768-203246768 on Build GRCh38
6353187 c.189G>C p.E63D Chr.2: 203246821-203246821 on Build GRCh38
325102 c.166G>T p.E56* Chr.2: 203246798-203246798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772699408 Chr.2: 203246751-203246751 on Build GRCh38
rs10601301 Chr.2: 203246678-203246681 on Build GRCh38
rs767221383 Chr.2: 203246743-203246743 on Build GRCh38
rs1019481726 Chr.2: 203246775-203246775 on Build GRCh38
rs770412261 Chr.2: 203246724-203246724 on Build GRCh38
rs753330849 Chr.2: 203246763-203246763 on Build GRCh38
rs754972297 Chr.2: 203246768-203246768 on Build GRCh38
rs778089616 Chr.2: 203246800-203246800 on Build GRCh38
rs765814940 Chr.2: 203246758-203246758 on Build GRCh38
rs946985842 Chr.2: 203246696-203246696 on Build GRCh38
rs776064140 Chr.2: 203246728-203246728 on Build GRCh38
rs867642122 Chr.2: 203246713-203246717 on Build GRCh38
rs149848848 Chr.2: 203246718-203246722 on Build GRCh38
rs6435176 Chr.2: 203246720-203246720 on Build GRCh38
rs147992273 Chr.2: 203246754-203246754 on Build GRCh38
rs145879414 Chr.2: 203246783-203246783 on Build GRCh38
rs369581070 Chr.2: 203246721-203246721 on Build GRCh38
rs749070482 Chr.2: 203246675-203246676 on Build GRCh38
rs113887964 Chr.2: 203246679-203246679 on Build GRCh38
rs752590306 Chr.2: 203246774-203246774 on Build GRCh38
rs371877492 Chr.2: 203246752-203246752 on Build GRCh38
rs896785557 Chr.2: 203246685-203246685 on Build GRCh38
rs765123651 Chr.2: 203246770-203246770 on Build GRCh38
rs758083852 Chr.2: 203246785-203246785 on Build GRCh38
rs13015873 Chr.2: 203246671-203246671 on Build GRCh38
rs10586509 Chr.2: 203246676-203246679 on Build GRCh38
rs777838805 Chr.2: 203246707-203246707 on Build GRCh38
rs777283840 Chr.2: 203246722-203246722 on Build GRCh38
rs775813119 Chr.2: 203246762-203246762 on Build GRCh38
rs1052723785 Chr.2: 203246639-203246639 on Build GRCh38
rs763204325 Chr.2: 203246734-203246734 on Build GRCh38
rs772381747 Chr.2: 203246739-203246740 on Build GRCh38
rs760064056 Chr.2: 203246723-203246723 on Build GRCh38

View Full Product Details