Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118592654-118592915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923621 c.1074C>T p.D358D Chr.11: 118592798-118592798 on Build GRCh38
923620 c.1027C>G p.L343V Chr.11: 118592751-118592751 on Build GRCh38
5837943 c.1075_1076delGT p.V359fs*20 Chr.11: 118592799-118592800 on Build GRCh38
6602827 c.1075G>A p.V359I Chr.11: 118592799-118592799 on Build GRCh38
1746067 c.1081G>A p.V361M Chr.11: 118592805-118592805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551779919 Chr.11: 118592831-118592831 on Build GRCh38
rs782319185 Chr.11: 118592744-118592744 on Build GRCh38
rs782625582 Chr.11: 118592706-118592706 on Build GRCh38
rs782151169 Chr.11: 118592791-118592791 on Build GRCh38
rs782256628 Chr.11: 118592686-118592686 on Build GRCh38
rs369324137 Chr.11: 118592829-118592829 on Build GRCh38
rs782813978 Chr.11: 118592793-118592793 on Build GRCh38
rs201898520 Chr.11: 118592798-118592798 on Build GRCh38
rs782574728 Chr.11: 118592837-118592837 on Build GRCh38
rs782369486 Chr.11: 118592766-118592766 on Build GRCh38
rs782180226 Chr.11: 118592721-118592721 on Build GRCh38
rs782258280 Chr.11: 118592866-118592866 on Build GRCh38
rs782789244 Chr.11: 118592835-118592835 on Build GRCh38
rs781832837 Chr.11: 118592804-118592804 on Build GRCh38
rs782103705 Chr.11: 118592771-118592771 on Build GRCh38
rs781926393 Chr.11: 118592873-118592873 on Build GRCh38
rs190216359 Chr.11: 118592685-118592685 on Build GRCh38
rs782334407 Chr.11: 118592689-118592689 on Build GRCh38
rs782412960 Chr.11: 118592863-118592863 on Build GRCh38
rs782274425 Chr.11: 118592839-118592839 on Build GRCh38
rs376911520 Chr.11: 118592778-118592778 on Build GRCh38
rs200207376 Chr.11: 118592754-118592754 on Build GRCh38
rs781956230 Chr.11: 118592768-118592768 on Build GRCh38
rs782524829 Chr.11: 118592799-118592799 on Build GRCh38
rs373229351 Chr.11: 118592838-118592838 on Build GRCh38
rs373132242 Chr.11: 118592753-118592753 on Build GRCh38
rs782162191 Chr.11: 118592765-118592765 on Build GRCh38
rs781811043 Chr.11: 118592789-118592789 on Build GRCh38
rs782069865 Chr.11: 118592875-118592875 on Build GRCh38
rs569999659 Chr.11: 118592868-118592868 on Build GRCh38
rs917545313 Chr.11: 118592872-118592872 on Build GRCh38

View Full Product Details