Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27289653-27289910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EQTN
Entrez Gene ID
54586
Gene Name
equatorin
Gene Aliases
AFAF, C9orf11, SPACA8
UniGene
Hs.163070
Gene Chromosome Location
Chr.9: 27284654-27297139 on Build GRCh38
Transcripts
NM_001161585.1 NM_020641.2 XM_011517920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6473116 c.433A>G p.T145A Chr.9: 27289720-27289720 on Build GRCh38
4626420 c.473C>T p.P158L Chr.9: 27289680-27289680 on Build GRCh38
1108010 c.426A>G p.I142M Chr.9: 27289727-27289727 on Build GRCh38
7291204 c.426A>C p.I142I Chr.9: 27289727-27289727 on Build GRCh38
346117 c.448G>T p.D150Y Chr.9: 27289705-27289705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764122930 Chr.9: 27289721-27289721 on Build GRCh38
rs752862310 Chr.9: 27289713-27289713 on Build GRCh38
rs544842895 Chr.9: 27289765-27289765 on Build GRCh38
rs769670845 Chr.9: 27289683-27289683 on Build GRCh38
rs955183084 Chr.9: 27289744-27289744 on Build GRCh38
rs114864861 Chr.9: 27289821-27289821 on Build GRCh38
rs575640951 Chr.9: 27289817-27289817 on Build GRCh38
rs780960420 Chr.9: 27289729-27289729 on Build GRCh38
rs774027210 Chr.9: 27289698-27289698 on Build GRCh38
rs776988695 Chr.9: 27289771-27289771 on Build GRCh38
rs759812649 Chr.9: 27289707-27289707 on Build GRCh38
rs981327242 Chr.9: 27289843-27289843 on Build GRCh38
rs775279057 Chr.9: 27289686-27289686 on Build GRCh38
rs1980660 Chr.9: 27289834-27289834 on Build GRCh38
rs915851636 Chr.9: 27289823-27289823 on Build GRCh38
rs540324269 Chr.9: 27289850-27289850 on Build GRCh38
rs770081432 Chr.9: 27289780-27289780 on Build GRCh38
rs768444112 Chr.9: 27289757-27289757 on Build GRCh38
rs928030954 Chr.9: 27289844-27289844 on Build GRCh38
rs367578113 Chr.9: 27289703-27289703 on Build GRCh38
rs765688896 Chr.9: 27289709-27289709 on Build GRCh38
rs369823902 Chr.9: 27289838-27289838 on Build GRCh38
rs137984899 Chr.9: 27289702-27289702 on Build GRCh38
rs771675097 Chr.9: 27289770-27289770 on Build GRCh38
rs200922368 Chr.9: 27289741-27289741 on Build GRCh38
rs907527371 Chr.9: 27289796-27289796 on Build GRCh38
rs932794574 Chr.9: 27289851-27289851 on Build GRCh38
rs187485851 Chr.9: 27289687-27289687 on Build GRCh38
rs374636857 Chr.9: 27289865-27289865 on Build GRCh38
rs778756398 Chr.9: 27289764-27289764 on Build GRCh38
rs751664261 Chr.9: 27289725-27289725 on Build GRCh38
rs762015177 Chr.9: 27289774-27289774 on Build GRCh38
rs192858288 Chr.9: 27289845-27289845 on Build GRCh38
rs534342710 Chr.9: 27289717-27289717 on Build GRCh38
rs757407388 Chr.9: 27289727-27289727 on Build GRCh38
rs768212994 Chr.9: 27289693-27289693 on Build GRCh38
rs760098490 Chr.9: 27289776-27289776 on Build GRCh38
rs1980661 Chr.9: 27289814-27289814 on Build GRCh38
rs748907309 Chr.9: 27289747-27289747 on Build GRCh38
rs558735779 Chr.9: 27289734-27289734 on Build GRCh38
rs143541625 Chr.9: 27289732-27289732 on Build GRCh38

View Full Product Details