Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102862031-102862302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS7
Entrez Gene ID
119032
Gene Name
BLOC-1 related complex subunit 7
Gene Aliases
C10orf32
UniGene
Hs.34492
Gene Chromosome Location
Chr.10: 102854210-102864961 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

SNP Detail

SNP ID Chromosome Location
rs773518393 Chr.10: 102862116-102862116 on Build GRCh38
rs370130274 Chr.10: 102862173-102862173 on Build GRCh38
rs1030490073 Chr.10: 102862123-102862123 on Build GRCh38
rs200985701 Chr.10: 102862210-102862210 on Build GRCh38
rs768444845 Chr.10: 102862215-102862215 on Build GRCh38
rs895885476 Chr.10: 102862246-102862246 on Build GRCh38
rs774302782 Chr.10: 102862223-102862223 on Build GRCh38
rs748056662 Chr.10: 102862224-102862224 on Build GRCh38
rs1057322102 Chr.10: 102862160-102862160 on Build GRCh38
rs900946175 Chr.10: 102862234-102862234 on Build GRCh38
rs755528543 Chr.10: 102862133-102862133 on Build GRCh38
rs941880319 Chr.10: 102862191-102862191 on Build GRCh38
rs765824088 Chr.10: 102862141-102862141 on Build GRCh38
rs956342602 Chr.10: 102862087-102862087 on Build GRCh38
rs973269048 Chr.10: 102862203-102862203 on Build GRCh38
rs145804818 Chr.10: 102862093-102862093 on Build GRCh38
rs41287472 Chr.10: 102862072-102862072 on Build GRCh38
rs1031947274 Chr.10: 102862085-102862085 on Build GRCh38
rs948792873 Chr.10: 102862258-102862258 on Build GRCh38
rs745469851 Chr.10: 102862188-102862188 on Build GRCh38
rs755771289 Chr.10: 102862192-102862192 on Build GRCh38
rs748841116 Chr.10: 102862213-102862213 on Build GRCh38
rs563084503 Chr.10: 102862079-102862079 on Build GRCh38
rs766498697 Chr.10: 102862128-102862128 on Build GRCh38
rs375952737 Chr.10: 102862230-102862230 on Build GRCh38
rs1015255867 Chr.10: 102862233-102862233 on Build GRCh38
rs377058545 Chr.10: 102862130-102862130 on Build GRCh38
rs1013055046 Chr.10: 102862197-102862197 on Build GRCh38
rs781176148 Chr.10: 102862116-102862117 on Build GRCh38
rs919197972 Chr.10: 102862205-102862205 on Build GRCh38
rs199954349 Chr.10: 102862152-102862152 on Build GRCh38
rs752974024 Chr.10: 102862145-102862145 on Build GRCh38
rs201568595 Chr.10: 102862156-102862156 on Build GRCh38
rs372982070 Chr.10: 102862178-102862178 on Build GRCh38
rs760988646 Chr.10: 102862124-102862124 on Build GRCh38

View Full Product Details