Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48765776-48766025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD5
Entrez Gene ID
392465
Gene Name
glyoxalase domain containing 5
Gene Aliases
-
UniGene
Hs.437539
Gene Chromosome Location
Chr.X: 48761572-48773648 on Build GRCh38
Transcripts
NM_001080489.2 XM_011543911.2 XM_011543912.2

SNP Detail

SNP ID Chromosome Location
rs371833787 Chr.X: 48765878-48765878 on Build GRCh38
rs782809626 Chr.X: 48765895-48765895 on Build GRCh38
rs782368125 Chr.X: 48765881-48765881 on Build GRCh38
rs782581497 Chr.X: 48765965-48765965 on Build GRCh38
rs782256259 Chr.X: 48765841-48765841 on Build GRCh38
rs782512630 Chr.X: 48765919-48765919 on Build GRCh38
rs781817808 Chr.X: 48765961-48765961 on Build GRCh38
rs374435425 Chr.X: 48765838-48765838 on Build GRCh38
rs368403511 Chr.X: 48765839-48765839 on Build GRCh38
rs781915112 Chr.X: 48765897-48765897 on Build GRCh38
rs782559282 Chr.X: 48765903-48765903 on Build GRCh38
rs782053658 Chr.X: 48765868-48765868 on Build GRCh38
rs368829297 Chr.X: 48765831-48765831 on Build GRCh38
rs782000639 Chr.X: 48765888-48765888 on Build GRCh38
rs782400252 Chr.X: 48765842-48765842 on Build GRCh38
rs782672176 Chr.X: 48765848-48765848 on Build GRCh38
rs782746948 Chr.X: 48765902-48765902 on Build GRCh38
rs782555536 Chr.X: 48765894-48765894 on Build GRCh38
rs267606469 Chr.X: 48765876-48765876 on Build GRCh38
rs199673981 Chr.X: 48765898-48765898 on Build GRCh38
rs781892463 Chr.X: 48765955-48765955 on Build GRCh38
rs200192031 Chr.X: 48765952-48765952 on Build GRCh38
rs782283794 Chr.X: 48765817-48765817 on Build GRCh38
rs984922419 Chr.X: 48765926-48765926 on Build GRCh38
rs781972442 Chr.X: 48765856-48765856 on Build GRCh38
rs956893453 Chr.X: 48765814-48765814 on Build GRCh38
rs143505421 Chr.X: 48765815-48765815 on Build GRCh38
rs782721563 Chr.X: 48765877-48765877 on Build GRCh38
rs782133429 Chr.X: 48765807-48765807 on Build GRCh38

View Full Product Details