Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15517041-15517300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA8
Entrez Gene ID
8516
Gene Name
integrin subunit alpha 8
Gene Aliases
-
UniGene
Hs.171311
Gene Chromosome Location
Chr.10: 15513949-15720335 on Build GRCh38
Transcripts
NM_001291494.1 NM_003638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3436219 c.3156G>A p.R1052R Chr.10: 15517194-15517194 on Build GRCh38
7372590 c.3182C>A p.P1061H Chr.10: 15517168-15517168 on Build GRCh38
6129084 c.3155G>C p.R1052T Chr.10: 15517195-15517195 on Build GRCh38
5932628 c.3133C>T p.P1045S Chr.10: 15517217-15517217 on Build GRCh38
4929582 c.3179C>G p.T1060S Chr.10: 15517171-15517171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371064779 Chr.10: 15517187-15517187 on Build GRCh38
rs550092776 Chr.10: 15517155-15517155 on Build GRCh38
rs564278369 Chr.10: 15517062-15517062 on Build GRCh38
rs199977895 Chr.10: 15517200-15517200 on Build GRCh38
rs758320081 Chr.10: 15517179-15517179 on Build GRCh38
rs773555839 Chr.10: 15517246-15517246 on Build GRCh38
rs773611380 Chr.10: 15517152-15517152 on Build GRCh38
rs561911227 Chr.10: 15517199-15517199 on Build GRCh38
rs531468585 Chr.10: 15517075-15517075 on Build GRCh38
rs762341278 Chr.10: 15517116-15517116 on Build GRCh38
rs754366716 Chr.10: 15517144-15517145 on Build GRCh38
rs765237923 Chr.10: 15517169-15517169 on Build GRCh38
rs527255216 Chr.10: 15517145-15517145 on Build GRCh38
rs9333252 Chr.10: 15517145-15517146 on Build GRCh38
rs539547678 Chr.10: 15517222-15517222 on Build GRCh38
rs772517596 Chr.10: 15517147-15517147 on Build GRCh38
rs1036696395 Chr.10: 15517070-15517070 on Build GRCh38
rs374089498 Chr.10: 15517280-15517280 on Build GRCh38
rs756960220 Chr.10: 15517194-15517194 on Build GRCh38
rs766472273 Chr.10: 15517140-15517142 on Build GRCh38
rs776988253 Chr.10: 15517271-15517271 on Build GRCh38
rs762985188 Chr.10: 15517276-15517276 on Build GRCh38
rs201629498 Chr.10: 15517219-15517219 on Build GRCh38
rs765501898 Chr.10: 15517275-15517275 on Build GRCh38
rs936037446 Chr.10: 15517073-15517073 on Build GRCh38
rs142568425 Chr.10: 15517161-15517161 on Build GRCh38
rs547401440 Chr.10: 15517217-15517217 on Build GRCh38
rs201573051 Chr.10: 15517218-15517218 on Build GRCh38
rs898281955 Chr.10: 15517160-15517160 on Build GRCh38
rs759750390 Chr.10: 15517273-15517273 on Build GRCh38
rs551516857 Chr.10: 15517267-15517267 on Build GRCh38
rs761269484 Chr.10: 15517253-15517253 on Build GRCh38
rs759772541 Chr.10: 15517167-15517167 on Build GRCh38
rs766800568 Chr.10: 15517159-15517159 on Build GRCh38
rs752842053 Chr.10: 15517178-15517178 on Build GRCh38
rs889393423 Chr.10: 15517088-15517088 on Build GRCh38
rs765778643 Chr.10: 15517211-15517211 on Build GRCh38

View Full Product Details