Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219968753-219969010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPRS
Entrez Gene ID
2058
Gene Name
glutamyl-prolyl-tRNA synthetase
Gene Aliases
EARS, GLUPRORS, PARS, PIG32, QARS, QPRS
UniGene
Hs.497788
Gene Chromosome Location
Chr.1: 219968600-220046658 on Build GRCh38
Transcripts
NM_004446.2 XM_017000614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3966276 c.4425A>T p.G1475G Chr.1: 219968920-219968920 on Build GRCh38
392620 c.4424delG p.G1475fs*>38 Chr.1: 219968921-219968921 on Build GRCh38
6534239 c.4411G>C p.A1471P Chr.1: 219968934-219968934 on Build GRCh38
5934286 c.4505C>T p.A1502V Chr.1: 219968840-219968840 on Build GRCh38
5950189 c.4529G>A p.R1510H Chr.1: 219968816-219968816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372808567 Chr.1: 219968804-219968804 on Build GRCh38
rs763800046 Chr.1: 219968913-219968913 on Build GRCh38
rs772190916 Chr.1: 219968832-219968832 on Build GRCh38
rs758572151 Chr.1: 219968952-219968952 on Build GRCh38
rs754404436 Chr.1: 219968929-219968929 on Build GRCh38
rs767777855 Chr.1: 219968827-219968827 on Build GRCh38
rs547115453 Chr.1: 219968817-219968817 on Build GRCh38
rs745803409 Chr.1: 219968967-219968967 on Build GRCh38
rs781321466 Chr.1: 219968873-219968873 on Build GRCh38
rs748875081 Chr.1: 219968976-219968976 on Build GRCh38
rs768305350 Chr.1: 219968894-219968894 on Build GRCh38
rs781074422 Chr.1: 219968958-219968958 on Build GRCh38
rs1052741539 Chr.1: 219968936-219968936 on Build GRCh38
rs930995541 Chr.1: 219968901-219968901 on Build GRCh38
rs374476794 Chr.1: 219968828-219968828 on Build GRCh38
rs1002607835 Chr.1: 219968942-219968942 on Build GRCh38
rs766281610 Chr.1: 219968875-219968875 on Build GRCh38
rs759981104 Chr.1: 219968935-219968935 on Build GRCh38
rs532343740 Chr.1: 219968955-219968955 on Build GRCh38
rs758584091 Chr.1: 219968863-219968863 on Build GRCh38
rs765309419 Chr.1: 219968937-219968937 on Build GRCh38
rs773848407 Chr.1: 219968899-219968899 on Build GRCh38
rs770863251 Chr.1: 219968788-219968788 on Build GRCh38
rs775518538 Chr.1: 219968839-219968839 on Build GRCh38
rs770827350 Chr.1: 219968870-219968870 on Build GRCh38
rs776303537 Chr.1: 219968789-219968789 on Build GRCh38
rs199545183 Chr.1: 219968889-219968889 on Build GRCh38
rs777464517 Chr.1: 219968868-219968868 on Build GRCh38
rs186287404 Chr.1: 219968840-219968840 on Build GRCh38
rs760274738 Chr.1: 219968904-219968904 on Build GRCh38
rs752786134 Chr.1: 219968951-219968951 on Build GRCh38
rs764426322 Chr.1: 219968954-219968954 on Build GRCh38
rs886842952 Chr.1: 219968931-219968931 on Build GRCh38
rs779451491 Chr.1: 219968974-219968974 on Build GRCh38
rs370238167 Chr.1: 219968973-219968973 on Build GRCh38
rs775338533 Chr.1: 219968809-219968809 on Build GRCh38
rs754310485 Chr.1: 219968835-219968835 on Build GRCh38
rs778954915 Chr.1: 219968857-219968857 on Build GRCh38
rs35055527 Chr.1: 219968904-219968904 on Build GRCh38
rs766916195 Chr.1: 219968834-219968834 on Build GRCh38
rs766721974 Chr.1: 219968905-219968905 on Build GRCh38
rs752878013 Chr.1: 219968860-219968860 on Build GRCh38
rs892388297 Chr.1: 219968961-219968961 on Build GRCh38
rs746928268 Chr.1: 219968787-219968787 on Build GRCh38
rs769440910 Chr.1: 219968874-219968874 on Build GRCh38
rs759167401 Chr.1: 219968797-219968797 on Build GRCh38
rs374384957 Chr.1: 219968977-219968977 on Build GRCh38
rs746770023 Chr.1: 219968869-219968869 on Build GRCh38
rs769398147 Chr.1: 219968799-219968799 on Build GRCh38
rs761104636 Chr.1: 219968830-219968830 on Build GRCh38

View Full Product Details