Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29017113-29017386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179A
Entrez Gene ID
165186
Gene Name
family with sequence similarity 179 member A
Gene Aliases
-
UniGene
Hs.525977
Gene Chromosome Location
Chr.2: 28981298-29052230 on Build GRCh38
Transcripts
NM_001321538.1 NM_001321539.1 NM_199280.2 XM_011532628.2 XM_011532631.1 XM_017003507.1 XM_017003508.1 XM_017003509.1 XM_017003510.1 XM_017003511.1 XM_017003512.1 XM_017003514.1 XM_017003516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437791 c.1157C>A p.S386Y Chr.2: 29017266-29017266 on Build GRCh38
1284831 c.1174C>T p.Q392* Chr.2: 29017283-29017283 on Build GRCh38
721302 c.1121G>T p.R374M Chr.2: 29017230-29017230 on Build GRCh38
7367184 c.1157C>T p.S386F Chr.2: 29017266-29017266 on Build GRCh38
3839379 c.1117C>T p.R373W Chr.2: 29017226-29017226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763440132 Chr.2: 29017174-29017174 on Build GRCh38
rs748488016 Chr.2: 29017193-29017193 on Build GRCh38
rs757946364 Chr.2: 29017272-29017272 on Build GRCh38
rs763883767 Chr.2: 29017259-29017259 on Build GRCh38
rs543539008 Chr.2: 29017181-29017181 on Build GRCh38
rs910822307 Chr.2: 29017246-29017246 on Build GRCh38
rs761410479 Chr.2: 29017194-29017217 on Build GRCh38
rs774471433 Chr.2: 29017223-29017223 on Build GRCh38
rs766384305 Chr.2: 29017145-29017145 on Build GRCh38
rs764766157 Chr.2: 29017203-29017211 on Build GRCh38
rs778152568 Chr.2: 29017216-29017216 on Build GRCh38
rs751163226 Chr.2: 29017282-29017282 on Build GRCh38
rs768749498 Chr.2: 29017337-29017337 on Build GRCh38
rs11127202 Chr.2: 29017194-29017194 on Build GRCh38
rs1033575893 Chr.2: 29017317-29017317 on Build GRCh38
rs779729483 Chr.2: 29017191-29017191 on Build GRCh38
rs1010028017 Chr.2: 29017273-29017273 on Build GRCh38
rs772222478 Chr.2: 29017227-29017227 on Build GRCh38
rs200034953 Chr.2: 29017345-29017345 on Build GRCh38
rs956827865 Chr.2: 29017185-29017185 on Build GRCh38
rs202125633 Chr.2: 29017268-29017268 on Build GRCh38
rs114252468 Chr.2: 29017357-29017357 on Build GRCh38
rs1041441895 Chr.2: 29017324-29017324 on Build GRCh38
rs763039115 Chr.2: 29017144-29017144 on Build GRCh38
rs765247449 Chr.2: 29017267-29017267 on Build GRCh38
rs866589416 Chr.2: 29017182-29017182 on Build GRCh38
rs776619466 Chr.2: 29017341-29017341 on Build GRCh38
rs779592142 Chr.2: 29017275-29017275 on Build GRCh38
rs755906864 Chr.2: 29017172-29017172 on Build GRCh38
rs779360500 Chr.2: 29017321-29017321 on Build GRCh38
rs201148338 Chr.2: 29017265-29017265 on Build GRCh38
rs1002475223 Chr.2: 29017332-29017332 on Build GRCh38
rs371456773 Chr.2: 29017325-29017325 on Build GRCh38
rs201395358 Chr.2: 29017219-29017219 on Build GRCh38
rs756437206 Chr.2: 29017289-29017289 on Build GRCh38
rs770247797 Chr.2: 29017138-29017138 on Build GRCh38
rs780776946 Chr.2: 29017336-29017336 on Build GRCh38
rs749638434 Chr.2: 29017295-29017295 on Build GRCh38
rs898204581 Chr.2: 29017279-29017279 on Build GRCh38
rs778635554 Chr.2: 29017178-29017178 on Build GRCh38
rs767765261 Chr.2: 29017159-29017159 on Build GRCh38
rs771422334 Chr.2: 29017221-29017221 on Build GRCh38
rs761673048 Chr.2: 29017342-29017342 on Build GRCh38
rs753751336 Chr.2: 29017177-29017177 on Build GRCh38
rs775580881 Chr.2: 29017331-29017331 on Build GRCh38
rs200945881 Chr.2: 29017226-29017226 on Build GRCh38
rs750332223 Chr.2: 29017353-29017353 on Build GRCh38
rs774557617 Chr.2: 29017153-29017153 on Build GRCh38
rs929719899 Chr.2: 29017310-29017310 on Build GRCh38
rs759819548 Chr.2: 29017157-29017157 on Build GRCh38
rs760401640 Chr.2: 29017251-29017251 on Build GRCh38
rs775733626 Chr.2: 29017240-29017240 on Build GRCh38
rs539972767 Chr.2: 29017239-29017239 on Build GRCh38
rs61730371 Chr.2: 29017184-29017184 on Build GRCh38
rs61750881 Chr.2: 29017266-29017266 on Build GRCh38
rs771475670 Chr.2: 29017314-29017314 on Build GRCh38
rs752483257 Chr.2: 29017171-29017171 on Build GRCh38
rs386644311 Chr.2: 29017265-29017266 on Build GRCh38
rs772132921 Chr.2: 29017327-29017327 on Build GRCh38
rs13409097 Chr.2: 29017139-29017139 on Build GRCh38

View Full Product Details