Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11649628-11649901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
UniGene
Hs.132753
Gene Chromosome Location
Chr.1: 11648361-11654831 on Build GRCh38
Transcripts
NM_012168.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620802 c.587C>T p.T196M Chr.1: 11649809-11649809 on Build GRCh38
462699 c.546T>G p.I182M Chr.1: 11649850-11649850 on Build GRCh38
5804344 c.554A>T p.Q185L Chr.1: 11649842-11649842 on Build GRCh38
333610 c.611A>T p.K204M Chr.1: 11649785-11649785 on Build GRCh38
6540094 c.559G>A p.E187K Chr.1: 11649837-11649837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373751221 Chr.1: 11649869-11649869 on Build GRCh38
rs75074481 Chr.1: 11649789-11649789 on Build GRCh38
rs2235658 Chr.1: 11649707-11649707 on Build GRCh38
rs748568075 Chr.1: 11649739-11649739 on Build GRCh38
rs373387055 Chr.1: 11649809-11649809 on Build GRCh38
rs745424628 Chr.1: 11649800-11649800 on Build GRCh38
rs762166327 Chr.1: 11649737-11649737 on Build GRCh38
rs751451284 Chr.1: 11649793-11649793 on Build GRCh38
rs149209829 Chr.1: 11649838-11649838 on Build GRCh38
rs759801772 Chr.1: 11649748-11649748 on Build GRCh38
rs926890074 Chr.1: 11649761-11649761 on Build GRCh38
rs951423334 Chr.1: 11649693-11649693 on Build GRCh38
rs769397284 Chr.1: 11649730-11649730 on Build GRCh38
rs765437441 Chr.1: 11649756-11649756 on Build GRCh38
rs758234107 Chr.1: 11649779-11649779 on Build GRCh38
rs35333217 Chr.1: 11649802-11649802 on Build GRCh38
rs527366614 Chr.1: 11649701-11649701 on Build GRCh38
rs550218124 Chr.1: 11649653-11649654 on Build GRCh38
rs780746164 Chr.1: 11649798-11649798 on Build GRCh38
rs759584337 Chr.1: 11649866-11649866 on Build GRCh38
rs377543023 Chr.1: 11649849-11649849 on Build GRCh38
rs772514172 Chr.1: 11649822-11649822 on Build GRCh38
rs1026179035 Chr.1: 11649682-11649682 on Build GRCh38
rs958029430 Chr.1: 11649708-11649708 on Build GRCh38
rs901797080 Chr.1: 11649651-11649651 on Build GRCh38
rs774762167 Chr.1: 11649733-11649733 on Build GRCh38
rs772193852 Chr.1: 11649779-11649780 on Build GRCh38
rs755765419 Chr.1: 11649806-11649806 on Build GRCh38
rs761284954 Chr.1: 11649741-11649741 on Build GRCh38
rs763821921 Chr.1: 11649878-11649878 on Build GRCh38
rs533202602 Chr.1: 11649652-11649652 on Build GRCh38
rs551599406 Chr.1: 11649663-11649663 on Build GRCh38
rs867521274 Chr.1: 11649671-11649671 on Build GRCh38
rs114834790 Chr.1: 11649835-11649835 on Build GRCh38
rs369806795 Chr.1: 11649744-11649744 on Build GRCh38
rs752514688 Chr.1: 11649762-11649762 on Build GRCh38
rs973576630 Chr.1: 11649697-11649697 on Build GRCh38
rs757187782 Chr.1: 11649795-11649795 on Build GRCh38
rs766506868 Chr.1: 11649742-11649742 on Build GRCh38
rs369589187 Chr.1: 11649867-11649867 on Build GRCh38
rs773814911 Chr.1: 11649740-11649740 on Build GRCh38
rs763911794 Chr.1: 11649792-11649792 on Build GRCh38
rs566473766 Chr.1: 11649694-11649694 on Build GRCh38
rs763186894 Chr.1: 11649874-11649874 on Build GRCh38
rs375861333 Chr.1: 11649879-11649879 on Build GRCh38
rs771408901 Chr.1: 11649847-11649847 on Build GRCh38

View Full Product Details