Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108376380-108376653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
UniGene
Hs.225968
Gene Chromosome Location
Chr.3: 108296490-108378285 on Build GRCh38
Transcripts
NM_001282556.1 NM_001282557.1 NM_001282558.1 NM_001282559.1 NM_007072.3 XM_005247080.3 XM_011512362.1 XM_011512363.1 XM_011512364.1 XM_011512367.2 XM_017005642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211464 c.1224+2T>A p.? Chr.3: 108376559-108376559 on Build GRCh38
2916784 c.1161A>G p.E387E Chr.3: 108376494-108376494 on Build GRCh38
5898822 c.1160-1G>A p.? Chr.3: 108376492-108376492 on Build GRCh38
6095835 c.1199C>T p.A400V Chr.3: 108376532-108376532 on Build GRCh38
4157118 c.1185G>A p.E395E Chr.3: 108376518-108376518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766170386 Chr.3: 108376536-108376536 on Build GRCh38
rs774834594 Chr.3: 108376443-108376443 on Build GRCh38
rs111477207 Chr.3: 108376477-108376477 on Build GRCh38
rs953281964 Chr.3: 108376591-108376591 on Build GRCh38
rs757649804 Chr.3: 108376499-108376499 on Build GRCh38
rs762903104 Chr.3: 108376544-108376544 on Build GRCh38
rs772986133 Chr.3: 108376529-108376529 on Build GRCh38
rs767920847 Chr.3: 108376565-108376565 on Build GRCh38
rs932188226 Chr.3: 108376430-108376430 on Build GRCh38
rs916945514 Chr.3: 108376480-108376480 on Build GRCh38
rs780381888 Chr.3: 108376585-108376585 on Build GRCh38
rs371340278 Chr.3: 108376513-108376513 on Build GRCh38
rs981355351 Chr.3: 108376470-108376470 on Build GRCh38
rs756502139 Chr.3: 108376569-108376569 on Build GRCh38
rs762821323 Chr.3: 108376475-108376477 on Build GRCh38
rs367804995 Chr.3: 108376487-108376487 on Build GRCh38
rs751741182 Chr.3: 108376546-108376546 on Build GRCh38
rs751354202 Chr.3: 108376477-108376477 on Build GRCh38
rs202188736 Chr.3: 108376465-108376465 on Build GRCh38
rs79390534 Chr.3: 108376566-108376566 on Build GRCh38
rs746287442 Chr.3: 108376510-108376510 on Build GRCh38
rs768241814 Chr.3: 108376520-108376520 on Build GRCh38
rs2124736 Chr.3: 108376518-108376518 on Build GRCh38
rs544693566 Chr.3: 108376521-108376521 on Build GRCh38
rs777909863 Chr.3: 108376557-108376557 on Build GRCh38
rs778362191 Chr.3: 108376620-108376620 on Build GRCh38
rs760497751 Chr.3: 108376475-108376475 on Build GRCh38
rs1039979198 Chr.3: 108376514-108376514 on Build GRCh38
rs968369602 Chr.3: 108376558-108376558 on Build GRCh38
rs776635433 Chr.3: 108376537-108376537 on Build GRCh38
rs760576388 Chr.3: 108376533-108376533 on Build GRCh38
rs929639826 Chr.3: 108376484-108376484 on Build GRCh38
rs374719479 Chr.3: 108376517-108376517 on Build GRCh38
rs184070601 Chr.3: 108376541-108376541 on Build GRCh38
rs755525786 Chr.3: 108376602-108376602 on Build GRCh38
rs987651873 Chr.3: 108376614-108376614 on Build GRCh38
rs773993803 Chr.3: 108376452-108376452 on Build GRCh38
rs749821837 Chr.3: 108376600-108376600 on Build GRCh38
rs764360113 Chr.3: 108376477-108376478 on Build GRCh38
rs374982963 Chr.3: 108376545-108376545 on Build GRCh38
rs761430147 Chr.3: 108376525-108376525 on Build GRCh38
rs371642037 Chr.3: 108376519-108376519 on Build GRCh38
rs761553106 Chr.3: 108376456-108376456 on Build GRCh38
rs750266073 Chr.3: 108376467-108376467 on Build GRCh38
rs77035676 Chr.3: 108376508-108376508 on Build GRCh38
rs771751375 Chr.3: 108376528-108376528 on Build GRCh38
rs573785338 Chr.3: 108376476-108376477 on Build GRCh38
rs899171379 Chr.3: 108376502-108376502 on Build GRCh38
rs751863436 Chr.3: 108376488-108376488 on Build GRCh38
rs752706157 Chr.3: 108376485-108376485 on Build GRCh38
rs768132102 Chr.3: 108376451-108376451 on Build GRCh38

View Full Product Details