Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 92590152-92590425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRID2
Entrez Gene ID
2895
Gene Name
glutamate ionotropic receptor delta type subunit 2
Gene Aliases
GluD2, SCAR18
UniGene
Hs.162727
Gene Chromosome Location
Chr.4: 92304249-93810330 on Build GRCh38
Transcripts
NM_001286838.1 NM_001510.3 XM_017008122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
70977 c.112A>C p.K38Q Chr.4: 92590154-92590154 on Build GRCh38
1130995 c.203C>T p.T68M Chr.4: 92590245-92590245 on Build GRCh38
3607038 c.197C>T p.S66L Chr.4: 92590239-92590239 on Build GRCh38
4688583 c.243_244insGGTAAGGT p.A82fs*8 Chr.4: 92590285-92590286 on Build GRCh38
228058 c.168G>T p.E56D Chr.4: 92590210-92590210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530036007 Chr.4: 92590370-92590370 on Build GRCh38
rs752627509 Chr.4: 92590196-92590196 on Build GRCh38
rs4142874 Chr.4: 92590399-92590399 on Build GRCh38
rs925062774 Chr.4: 92590183-92590183 on Build GRCh38
rs768112918 Chr.4: 92590383-92590383 on Build GRCh38
rs777650242 Chr.4: 92590305-92590305 on Build GRCh38
rs368889618 Chr.4: 92590254-92590254 on Build GRCh38
rs1029645404 Chr.4: 92590279-92590279 on Build GRCh38
rs191198939 Chr.4: 92590319-92590319 on Build GRCh38
rs763741990 Chr.4: 92590205-92590207 on Build GRCh38
rs904327762 Chr.4: 92590206-92590206 on Build GRCh38
rs746848077 Chr.4: 92590213-92590213 on Build GRCh38
rs777523518 Chr.4: 92590212-92590212 on Build GRCh38
rs745551453 Chr.4: 92590243-92590243 on Build GRCh38
rs1047512345 Chr.4: 92590360-92590360 on Build GRCh38
rs759869521 Chr.4: 92590275-92590275 on Build GRCh38
rs370608333 Chr.4: 92590369-92590369 on Build GRCh38
rs369776528 Chr.4: 92590312-92590312 on Build GRCh38
rs769178842 Chr.4: 92590244-92590244 on Build GRCh38
rs761070179 Chr.4: 92590256-92590256 on Build GRCh38
rs752825623 Chr.4: 92590291-92590291 on Build GRCh38
rs769531690 Chr.4: 92590337-92590337 on Build GRCh38
rs753940665 Chr.4: 92590267-92590267 on Build GRCh38
rs942735898 Chr.4: 92590311-92590311 on Build GRCh38
rs35156583 Chr.4: 92590236-92590237 on Build GRCh38
rs766779972 Chr.4: 92590259-92590259 on Build GRCh38
rs765218145 Chr.4: 92590282-92590282 on Build GRCh38
rs772742894 Chr.4: 92590249-92590249 on Build GRCh38
rs375407020 Chr.4: 92590308-92590308 on Build GRCh38
rs770746104 Chr.4: 92590216-92590216 on Build GRCh38
rs34144324 Chr.4: 92590245-92590245 on Build GRCh38
rs866612183 Chr.4: 92590338-92590338 on Build GRCh38
rs749040083 Chr.4: 92590389-92590389 on Build GRCh38
rs549686369 Chr.4: 92590374-92590374 on Build GRCh38
rs563942882 Chr.4: 92590197-92590197 on Build GRCh38
rs145606176 Chr.4: 92590219-92590219 on Build GRCh38
rs2870639 Chr.4: 92590330-92590330 on Build GRCh38
rs371408681 Chr.4: 92590298-92590298 on Build GRCh38
rs559895523 Chr.4: 92590246-92590246 on Build GRCh38
rs779054312 Chr.4: 92590182-92590182 on Build GRCh38
rs752626265 Chr.4: 92590266-92590266 on Build GRCh38
rs958434108 Chr.4: 92590333-92590333 on Build GRCh38
rs752040593 Chr.4: 92590208-92590208 on Build GRCh38

View Full Product Details