Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177229866-177230135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP2
Entrez Gene ID
57795
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
DBCCR1L2, FAM5B
UniGene
Hs.495918
Gene Chromosome Location
Chr.1: 177170922-177282422 on Build GRCh38
Transcripts
NM_021165.3 XM_005245379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1166121 c.200G>A p.R67H Chr.1: 177230076-177230076 on Build GRCh38
676835 c.14G>A p.C5Y Chr.1: 177229890-177229890 on Build GRCh38
6060007 c.192C>T p.P64P Chr.1: 177230068-177230068 on Build GRCh38
2094278 c.145G>A p.V49I Chr.1: 177230021-177230021 on Build GRCh38
4025456 c.42G>A p.P14P Chr.1: 177229918-177229918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757573324 Chr.1: 177229893-177229893 on Build GRCh38
rs777303846 Chr.1: 177230008-177230008 on Build GRCh38
rs757041919 Chr.1: 177230021-177230021 on Build GRCh38
rs138435872 Chr.1: 177230077-177230077 on Build GRCh38
rs16850984 Chr.1: 177230088-177230088 on Build GRCh38
rs920926796 Chr.1: 177230069-177230069 on Build GRCh38
rs775532892 Chr.1: 177229938-177229938 on Build GRCh38
rs558022486 Chr.1: 177230037-177230037 on Build GRCh38
rs986474947 Chr.1: 177230102-177230102 on Build GRCh38
rs775336666 Chr.1: 177230051-177230051 on Build GRCh38
rs756669472 Chr.1: 177230003-177230003 on Build GRCh38
rs912146352 Chr.1: 177229987-177229987 on Build GRCh38
rs768655368 Chr.1: 177230111-177230111 on Build GRCh38
rs147833227 Chr.1: 177229885-177229885 on Build GRCh38
rs377272745 Chr.1: 177230044-177230044 on Build GRCh38
rs746624639 Chr.1: 177230020-177230020 on Build GRCh38
rs755648048 Chr.1: 177229916-177229916 on Build GRCh38
rs140915160 Chr.1: 177229917-177229917 on Build GRCh38
rs535197966 Chr.1: 177229924-177229924 on Build GRCh38
rs920899184 Chr.1: 177229951-177229951 on Build GRCh38
rs139209134 Chr.1: 177230092-177230092 on Build GRCh38
rs553424360 Chr.1: 177230114-177230114 on Build GRCh38
rs267598199 Chr.1: 177229903-177229903 on Build GRCh38
rs112761040 Chr.1: 177230072-177230072 on Build GRCh38
rs767880163 Chr.1: 177229989-177229989 on Build GRCh38
rs756920458 Chr.1: 177230087-177230087 on Build GRCh38
rs147698296 Chr.1: 177229909-177229909 on Build GRCh38
rs924857789 Chr.1: 177229898-177229898 on Build GRCh38
rs569033840 Chr.1: 177229942-177229942 on Build GRCh38
rs777691956 Chr.1: 177229918-177229918 on Build GRCh38
rs745666848 Chr.1: 177230095-177230095 on Build GRCh38
rs752251952 Chr.1: 177230006-177230006 on Build GRCh38
rs748381466 Chr.1: 177230112-177230112 on Build GRCh38
rs750937304 Chr.1: 177229999-177229999 on Build GRCh38
rs149441185 Chr.1: 177230005-177230005 on Build GRCh38
rs768728535 Chr.1: 177230060-177230060 on Build GRCh38
rs764152225 Chr.1: 177230087-177230088 on Build GRCh38
rs146202383 Chr.1: 177230086-177230086 on Build GRCh38
rs376635257 Chr.1: 177230056-177230056 on Build GRCh38
rs986749782 Chr.1: 177229969-177229969 on Build GRCh38
rs200656202 Chr.1: 177230115-177230115 on Build GRCh38
rs773700754 Chr.1: 177230075-177230075 on Build GRCh38
rs763602017 Chr.1: 177230064-177230064 on Build GRCh38
rs762102096 Chr.1: 177229986-177229986 on Build GRCh38
rs769910100 Chr.1: 177229937-177229937 on Build GRCh38
rs200249029 Chr.1: 177229913-177229913 on Build GRCh38
rs199631180 Chr.1: 177229930-177229930 on Build GRCh38
rs150664972 Chr.1: 177230107-177230107 on Build GRCh38
rs953661445 Chr.1: 177229963-177229963 on Build GRCh38
rs751744392 Chr.1: 177229977-177229977 on Build GRCh38
rs759437536 Chr.1: 177229935-177229935 on Build GRCh38
rs758005507 Chr.1: 177230082-177230082 on Build GRCh38
rs962477177 Chr.1: 177230059-177230059 on Build GRCh38
rs774464143 Chr.1: 177230062-177230062 on Build GRCh38
rs763621067 Chr.1: 177230085-177230085 on Build GRCh38
rs1049073734 Chr.1: 177230050-177230050 on Build GRCh38
rs569593343 Chr.1: 177229907-177229907 on Build GRCh38
rs755843993 Chr.1: 177230105-177230105 on Build GRCh38
rs751983601 Chr.1: 177230078-177230078 on Build GRCh38
rs944890706 Chr.1: 177229990-177229990 on Build GRCh38
rs568417156 Chr.1: 177229921-177229921 on Build GRCh38
rs747854235 Chr.1: 177229914-177229914 on Build GRCh38
rs764224385 Chr.1: 177229958-177229958 on Build GRCh38
rs767825106 Chr.1: 177230065-177230065 on Build GRCh38
rs781018149 Chr.1: 177230036-177230036 on Build GRCh38

View Full Product Details