Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111736816-111737077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK4
Entrez Gene ID
9732
Gene Name
dedicator of cytokinesis 4
Gene Aliases
-
UniGene
Hs.654652
Gene Chromosome Location
Chr.7: 111726108-112206407 on Build GRCh38
Transcripts
NM_014705.3 XM_006716188.2 XM_006716189.2 XM_017012819.1 XM_017012820.1 XM_017012821.1 XM_017012822.1 XM_017012823.1 XM_017012824.1 XM_017012825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406235 c.5255C>T p.S1752F Chr.7: 111736931-111736931 on Build GRCh38
5406236 c.1913C>T p.S638F Chr.7: 111736931-111736931 on Build GRCh38
3877257 c.5209A>G p.N1737D Chr.7: 111736977-111736977 on Build GRCh38
4358869 c.1879G>A p.D627N Chr.7: 111736965-111736965 on Build GRCh38
4964691 c.1882G>A p.G628R Chr.7: 111736962-111736962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751183443 Chr.7: 111736988-111736988 on Build GRCh38
rs1033036628 Chr.7: 111736904-111736904 on Build GRCh38
rs761046979 Chr.7: 111736961-111736961 on Build GRCh38
rs1026006181 Chr.7: 111737005-111737005 on Build GRCh38
rs768706438 Chr.7: 111736950-111736950 on Build GRCh38
rs757910835 Chr.7: 111736970-111736970 on Build GRCh38
rs953292573 Chr.7: 111737011-111737011 on Build GRCh38
rs755942259 Chr.7: 111737006-111737006 on Build GRCh38
rs775396975 Chr.7: 111736924-111736924 on Build GRCh38
rs762240664 Chr.7: 111736917-111736918 on Build GRCh38
rs773731165 Chr.7: 111736959-111736959 on Build GRCh38
rs184672508 Chr.7: 111736877-111736877 on Build GRCh38
rs202207944 Chr.7: 111736963-111736963 on Build GRCh38
rs763662318 Chr.7: 111736973-111736973 on Build GRCh38
rs535465340 Chr.7: 111736997-111736997 on Build GRCh38
rs375220879 Chr.7: 111737047-111737047 on Build GRCh38
rs189055113 Chr.7: 111737029-111737029 on Build GRCh38
rs149384765 Chr.7: 111736845-111736845 on Build GRCh38
rs1030665813 Chr.7: 111736865-111736865 on Build GRCh38
rs767870061 Chr.7: 111736995-111736996 on Build GRCh38
rs903154184 Chr.7: 111736871-111736871 on Build GRCh38
rs745783701 Chr.7: 111736883-111736883 on Build GRCh38
rs760442905 Chr.7: 111736847-111736847 on Build GRCh38
rs564183702 Chr.7: 111736949-111736949 on Build GRCh38
rs745710848 Chr.7: 111737001-111737001 on Build GRCh38
rs145638553 Chr.7: 111736848-111736848 on Build GRCh38
rs750781177 Chr.7: 111737005-111737006 on Build GRCh38
rs889027290 Chr.7: 111736937-111736937 on Build GRCh38
rs775902528 Chr.7: 111736895-111736895 on Build GRCh38
rs201048359 Chr.7: 111737030-111737030 on Build GRCh38
rs770246903 Chr.7: 111737044-111737044 on Build GRCh38
rs377250270 Chr.7: 111736854-111736854 on Build GRCh38
rs766914725 Chr.7: 111736962-111736962 on Build GRCh38

View Full Product Details