Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14515741-14516002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB1
Entrez Gene ID
3337
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Gene Aliases
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1
UniGene
Hs.515210
Gene Chromosome Location
Chr.19: 14514764-14529821 on Build GRCh38
Transcripts
NM_001300914.1 NM_001313964.1 NM_006145.2 XM_006722733.1 XM_006722734.3 XM_011527956.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647068 c.997G>A p.V333I Chr.19: 14515966-14515966 on Build GRCh38
6927888 c.1016C>T p.P339L Chr.19: 14515947-14515947 on Build GRCh38
6944819 c.1003G>A p.E335K Chr.19: 14515960-14515960 on Build GRCh38
7365045 c.983A>T p.Q328L Chr.19: 14515980-14515980 on Build GRCh38
4514557 c.979C>T p.P327S Chr.19: 14515984-14515984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775128939 Chr.19: 14515947-14515947 on Build GRCh38
rs779114840 Chr.19: 14515897-14515897 on Build GRCh38
rs377499028 Chr.19: 14515904-14515904 on Build GRCh38
rs552986746 Chr.19: 14515896-14515896 on Build GRCh38
rs945341670 Chr.19: 14515860-14515860 on Build GRCh38
rs910692895 Chr.19: 14515793-14515793 on Build GRCh38
rs762391144 Chr.19: 14515924-14515925 on Build GRCh38
rs553097812 Chr.19: 14515849-14515849 on Build GRCh38
rs775462084 Chr.19: 14515893-14515893 on Build GRCh38
rs768069872 Chr.19: 14515941-14515942 on Build GRCh38
rs183669246 Chr.19: 14515828-14515828 on Build GRCh38
rs753428745 Chr.19: 14515954-14515954 on Build GRCh38
rs958344132 Chr.19: 14515817-14515817 on Build GRCh38
rs774775077 Chr.19: 14515913-14515914 on Build GRCh38
rs138234955 Chr.19: 14515975-14515975 on Build GRCh38
rs534851944 Chr.19: 14515831-14515831 on Build GRCh38
rs532564939 Chr.19: 14515977-14515977 on Build GRCh38
rs755426483 Chr.19: 14515925-14515925 on Build GRCh38
rs541792196 Chr.19: 14515900-14515900 on Build GRCh38
rs774494870 Chr.19: 14515955-14515955 on Build GRCh38
rs767875619 Chr.19: 14515957-14515957 on Build GRCh38
rs202053180 Chr.19: 14515937-14515937 on Build GRCh38
rs769622044 Chr.19: 14515945-14515945 on Build GRCh38
rs561895564 Chr.19: 14515980-14515980 on Build GRCh38
rs762763836 Chr.19: 14515899-14515899 on Build GRCh38
rs954434270 Chr.19: 14515770-14515770 on Build GRCh38
rs928087570 Chr.19: 14515881-14515881 on Build GRCh38
rs1006152593 Chr.19: 14515908-14515908 on Build GRCh38
rs750830588 Chr.19: 14515913-14515913 on Build GRCh38
rs756381480 Chr.19: 14515966-14515966 on Build GRCh38
rs897501241 Chr.19: 14515924-14515924 on Build GRCh38
rs754996878 Chr.19: 14515863-14515863 on Build GRCh38
rs201154155 Chr.19: 14515956-14515956 on Build GRCh38
rs374890889 Chr.19: 14515934-14515934 on Build GRCh38
rs754487743 Chr.19: 14515971-14515971 on Build GRCh38
rs981319866 Chr.19: 14515853-14515853 on Build GRCh38
rs150137137 Chr.19: 14515954-14515954 on Build GRCh38
rs189216189 Chr.19: 14515921-14515921 on Build GRCh38
rs986351987 Chr.19: 14515791-14515791 on Build GRCh38
rs756497726 Chr.19: 14515915-14515915 on Build GRCh38
rs750634405 Chr.19: 14515965-14515965 on Build GRCh38
rs530905480 Chr.19: 14515982-14515982 on Build GRCh38
rs749848780 Chr.19: 14515923-14515923 on Build GRCh38
rs138322666 Chr.19: 14515867-14515867 on Build GRCh38
rs748281603 Chr.19: 14515907-14515907 on Build GRCh38
rs377601762 Chr.19: 14515928-14515928 on Build GRCh38
rs766824792 Chr.19: 14515967-14515967 on Build GRCh38

View Full Product Details