Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28527657-28527926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD3
Entrez Gene ID
7976
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3
UniGene
Hs.40735
Gene Chromosome Location
Chr.8: 28494205-28574268 on Build GRCh38
Transcripts
NM_017412.3 NM_145866.1 XM_017013841.1 XM_017013842.1 XM_017013843.1 XM_017013844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910227 c.1012A>G p.I338V Chr.8: 28527772-28527772 on Build GRCh38
4924345 c.1158G>C p.G386G Chr.8: 28527918-28527918 on Build GRCh38
4842395 c.1057G>T p.E353* Chr.8: 28527817-28527817 on Build GRCh38
1456492 c.1154T>C p.V385A Chr.8: 28527914-28527914 on Build GRCh38
750783 c.990G>A p.L330L Chr.8: 28527750-28527750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17059195 Chr.8: 28527777-28527777 on Build GRCh38
rs752661059 Chr.8: 28527802-28527802 on Build GRCh38
rs62001923 Chr.8: 28527827-28527827 on Build GRCh38
rs756222099 Chr.8: 28527803-28527803 on Build GRCh38
rs375741315 Chr.8: 28527838-28527838 on Build GRCh38
rs752082426 Chr.8: 28527788-28527788 on Build GRCh38
rs754930200 Chr.8: 28527858-28527858 on Build GRCh38
rs749229314 Chr.8: 28527814-28527814 on Build GRCh38
rs371916844 Chr.8: 28527837-28527837 on Build GRCh38
rs911191004 Chr.8: 28527721-28527721 on Build GRCh38
rs374350781 Chr.8: 28527688-28527688 on Build GRCh38
rs766846977 Chr.8: 28527878-28527878 on Build GRCh38
rs767844094 Chr.8: 28527890-28527890 on Build GRCh38
rs767693020 Chr.8: 28527790-28527790 on Build GRCh38
rs758164477 Chr.8: 28527807-28527807 on Build GRCh38
rs760651779 Chr.8: 28527733-28527733 on Build GRCh38
rs759992378 Chr.8: 28527889-28527889 on Build GRCh38
rs747574173 Chr.8: 28527847-28527847 on Build GRCh38
rs761404264 Chr.8: 28527758-28527758 on Build GRCh38
rs367706061 Chr.8: 28527805-28527805 on Build GRCh38
rs372290611 Chr.8: 28527859-28527859 on Build GRCh38
rs35150382 Chr.8: 28527693-28527694 on Build GRCh38
rs141884499 Chr.8: 28527759-28527759 on Build GRCh38
rs775355174 Chr.8: 28527708-28527708 on Build GRCh38
rs768120381 Chr.8: 28527744-28527744 on Build GRCh38
rs965485773 Chr.8: 28527809-28527809 on Build GRCh38
rs370665917 Chr.8: 28527685-28527685 on Build GRCh38
rs140023424 Chr.8: 28527879-28527879 on Build GRCh38
rs766901985 Chr.8: 28527782-28527782 on Build GRCh38
rs200938191 Chr.8: 28527756-28527756 on Build GRCh38
rs769370555 Chr.8: 28527856-28527856 on Build GRCh38
rs750124182 Chr.8: 28527765-28527765 on Build GRCh38
rs148953574 Chr.8: 28527804-28527804 on Build GRCh38

View Full Product Details