Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151468793-151469018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMT1
Entrez Gene ID
79624
Gene Name
acidic residue methyltransferase 1
Gene Aliases
C6orf211
UniGene
Hs.15929
Gene Chromosome Location
Chr.6: 151452258-151470101 on Build GRCh38
Transcripts
NM_001286562.1 NM_024573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3019390 c.1057T>C p.Y353H Chr.6: 151468841-151468841 on Build GRCh38
5881264 c.1187C>G p.P396R Chr.6: 151468971-151468971 on Build GRCh38
4917799 c.1159C>G p.Q387E Chr.6: 151468943-151468943 on Build GRCh38
4520754 c.1074G>A p.K358K Chr.6: 151468858-151468858 on Build GRCh38
3941554 c.1231C>T p.L411L Chr.6: 151469015-151469015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766688917 Chr.6: 151468973-151468973 on Build GRCh38
rs147127681 Chr.6: 151468817-151468817 on Build GRCh38
rs780753822 Chr.6: 151468997-151468997 on Build GRCh38
rs779458832 Chr.6: 151468984-151468984 on Build GRCh38
rs558614819 Chr.6: 151468878-151468878 on Build GRCh38
rs756757031 Chr.6: 151468991-151468991 on Build GRCh38
rs767698565 Chr.6: 151468968-151468968 on Build GRCh38
rs748184329 Chr.6: 151468879-151468879 on Build GRCh38
rs755419809 Chr.6: 151468978-151468978 on Build GRCh38
rs754347404 Chr.6: 151468974-151468974 on Build GRCh38
rs1024374299 Chr.6: 151468923-151468923 on Build GRCh38
rs201674567 Chr.6: 151468970-151468970 on Build GRCh38
rs140057628 Chr.6: 151468967-151468967 on Build GRCh38
rs774240808 Chr.6: 151468964-151468964 on Build GRCh38
rs757933424 Chr.6: 151468830-151468830 on Build GRCh38
rs892818372 Chr.6: 151468956-151468956 on Build GRCh38
rs372873070 Chr.6: 151468982-151468982 on Build GRCh38
rs988836237 Chr.6: 151468921-151468921 on Build GRCh38
rs140306203 Chr.6: 151468841-151468841 on Build GRCh38
rs770544009 Chr.6: 151468874-151468874 on Build GRCh38
rs775593762 Chr.6: 151468948-151468948 on Build GRCh38
rs151335520 Chr.6: 151468868-151468868 on Build GRCh38
rs762946533 Chr.6: 151468949-151468949 on Build GRCh38
rs374663926 Chr.6: 151468879-151468879 on Build GRCh38
rs971486366 Chr.6: 151468926-151468926 on Build GRCh38
rs754292482 Chr.6: 151468825-151468825 on Build GRCh38
rs780580999 Chr.6: 151468824-151468824 on Build GRCh38
rs149658464 Chr.6: 151468990-151468990 on Build GRCh38
rs142746071 Chr.6: 151468936-151468936 on Build GRCh38
rs140681772 Chr.6: 151468961-151468961 on Build GRCh38

View Full Product Details