Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63987878-63988151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
UniGene
Hs.476595
Gene Chromosome Location
Chr.3: 63864557-64003462 on Build GRCh38
Transcripts
NM_000333.3 NM_001128149.2 NM_001177387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664722 c.1113G>A p.Q371Q Chr.3: 63988076-63988076 on Build GRCh38
4459104 c.1111C>T p.Q371* Chr.3: 63988074-63988074 on Build GRCh38
2783380 c.1161C>T p.A387A Chr.3: 63988124-63988124 on Build GRCh38
2783384 c.1174A>G p.K392E Chr.3: 63988137-63988137 on Build GRCh38
4664721 c.1113G>A p.Q371Q Chr.3: 63988076-63988076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973084377 Chr.3: 63988091-63988091 on Build GRCh38
rs757330737 Chr.3: 63988090-63988090 on Build GRCh38
rs556049203 Chr.3: 63988105-63988105 on Build GRCh38
rs761799687 Chr.3: 63988043-63988043 on Build GRCh38
rs772535779 Chr.3: 63988050-63988050 on Build GRCh38
rs757673995 Chr.3: 63988042-63988043 on Build GRCh38
rs893478344 Chr.3: 63987939-63987939 on Build GRCh38
rs774397773 Chr.3: 63988039-63988039 on Build GRCh38
rs1029527524 Chr.3: 63988065-63988065 on Build GRCh38
rs1033754308 Chr.3: 63987933-63987933 on Build GRCh38
rs765549180 Chr.3: 63988106-63988106 on Build GRCh38
rs753083444 Chr.3: 63988113-63988113 on Build GRCh38
rs777438849 Chr.3: 63987958-63987958 on Build GRCh38
rs1031927945 Chr.3: 63987973-63987973 on Build GRCh38
rs368534595 Chr.3: 63988012-63988012 on Build GRCh38
rs778348924 Chr.3: 63988124-63988124 on Build GRCh38
rs187168095 Chr.3: 63987977-63987977 on Build GRCh38
rs895558696 Chr.3: 63987998-63987998 on Build GRCh38
rs72876990 Chr.3: 63988064-63988064 on Build GRCh38
rs746877558 Chr.3: 63988034-63988034 on Build GRCh38
rs777666953 Chr.3: 63988008-63988008 on Build GRCh38
rs548146108 Chr.3: 63987981-63987981 on Build GRCh38
rs954092116 Chr.3: 63988126-63988126 on Build GRCh38
rs114044696 Chr.3: 63987907-63987907 on Build GRCh38
rs776979597 Chr.3: 63988025-63988025 on Build GRCh38
rs767509914 Chr.3: 63988049-63988049 on Build GRCh38
rs762828259 Chr.3: 63988026-63988026 on Build GRCh38
rs111806673 Chr.3: 63988109-63988109 on Build GRCh38
rs765938407 Chr.3: 63988079-63988079 on Build GRCh38
rs770991872 Chr.3: 63988021-63988021 on Build GRCh38
rs182519316 Chr.3: 63987929-63987929 on Build GRCh38
rs952949407 Chr.3: 63988123-63988123 on Build GRCh38

View Full Product Details