Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55868369-55868608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
UniGene
Hs.269665
Gene Chromosome Location
Chr.4: 55853616-55905078 on Build GRCh38
Transcripts
NM_001024924.1 NM_018261.3 NM_178237.2 XM_005265747.2 XM_005265748.1 XM_005265750.2 XM_017008405.1 XM_017008406.1 XM_017008407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3974949 c.568G>T p.E190* Chr.4: 55868488-55868488 on Build GRCh38
4005777 c.497G>C p.R166T Chr.4: 55868417-55868417 on Build GRCh38
1056188 c.559G>A p.A187T Chr.4: 55868479-55868479 on Build GRCh38
6657081 c.581G>A p.R194K Chr.4: 55868501-55868501 on Build GRCh38
734113 c.493G>T p.A165S Chr.4: 55868413-55868413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758860475 Chr.4: 55868561-55868561 on Build GRCh38
rs780857976 Chr.4: 55868425-55868425 on Build GRCh38
rs748600001 Chr.4: 55868465-55868465 on Build GRCh38
rs763222605 Chr.4: 55868478-55868478 on Build GRCh38
rs779644856 Chr.4: 55868441-55868441 on Build GRCh38
rs778291574 Chr.4: 55868557-55868557 on Build GRCh38
rs773470317 Chr.4: 55868474-55868474 on Build GRCh38
rs3213948 Chr.4: 55868535-55868535 on Build GRCh38
rs200864049 Chr.4: 55868453-55868453 on Build GRCh38
rs1031050876 Chr.4: 55868428-55868428 on Build GRCh38
rs750179198 Chr.4: 55868433-55868433 on Build GRCh38
rs199645913 Chr.4: 55868494-55868494 on Build GRCh38
rs778310359 Chr.4: 55868564-55868564 on Build GRCh38
rs761674429 Chr.4: 55868508-55868508 on Build GRCh38
rs753478114 Chr.4: 55868543-55868543 on Build GRCh38
rs769307911 Chr.4: 55868501-55868501 on Build GRCh38
rs772571818 Chr.4: 55868468-55868468 on Build GRCh38
rs767444415 Chr.4: 55868509-55868509 on Build GRCh38
rs755875746 Chr.4: 55868533-55868533 on Build GRCh38
rs759722317 Chr.4: 55868521-55868521 on Build GRCh38
rs979991122 Chr.4: 55868498-55868498 on Build GRCh38
rs753426467 Chr.4: 55868463-55868463 on Build GRCh38
rs754550230 Chr.4: 55868544-55868544 on Build GRCh38
rs146961843 Chr.4: 55868475-55868475 on Build GRCh38
rs560881722 Chr.4: 55868574-55868574 on Build GRCh38
rs201297607 Chr.4: 55868487-55868487 on Build GRCh38
rs768134662 Chr.4: 55868459-55868459 on Build GRCh38
rs747644671 Chr.4: 55868559-55868559 on Build GRCh38
rs776968740 Chr.4: 55868491-55868491 on Build GRCh38
rs771317758 Chr.4: 55868480-55868480 on Build GRCh38
rs755789256 Chr.4: 55868434-55868434 on Build GRCh38
rs774712061 Chr.4: 55868418-55868420 on Build GRCh38
rs141291877 Chr.4: 55868444-55868444 on Build GRCh38
rs574805859 Chr.4: 55868499-55868499 on Build GRCh38

View Full Product Details